EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
LC-MS analysis of the plasma metabolome--a novel sample preparation strategy.
PMID 25531874
Abstract
Blood plasma is a well-known body fluid often analyzed in studies on the effects of toxic compounds as physiological or chemical induced changes in the mammalian body are reflected in the plasma metabolome. Sample preparation prior to LC-MS based analysis of the plasma metabolome is a challenge as plasma contains compounds with very different properties. Besides, proteins, which usually are precipitated with organic solvent, phospholipids, are known to cause ion suppression in electrospray mass spectrometry. We have compared two different sample preparation techniques prior to LC-qTOF analysis of plasma samples: the first is protein precipitation; the second is protein precipitation followed by solid phase extraction with sub-fractionation into three sub-samples: a phospholipid, a lipid and a polar sub-fraction. Molecular feature extraction of the data files from LC-qTOF analysis of the samples revealed 1792 molecular features from the protein precipitation procedure. The protein precipitation followed by solid phase extraction procedure with three sub-samples gave a total of 4234 molecular features. This suggests that sub-sampling into polar, lipid and phospholipid fractions enables extraction of more metabolomic information as compared to protein precipitation alone. Chromatography showed good separation of the metabolites with little retention time drift (<1s) and a mass accuracy below 3 ppm was observed. The performance of the method was investigated using plasma samples from rats administered the environmental pollutant perfluorononanoic acid.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
78551
3-(4-Methylbenzylidene)camphor, ≥98.0% (GC)
C18H22O
61547
3-(4-Methylbenzylidene)camphor, certified reference material, TraceCERT®
C18H22O
442840
4,4′-Isopropylidenediphenol, analytical standard
C15H16O2
67261
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®
C24H38O4
36735
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
C24H38O4
80030
Bis(2-ethylhexyl) phthalate, Selectophore
C24H38O4
239658
Bisphenol A, ≥99%
C15H16O2
42088
Bisphenol A, certified reference material, TraceCERT®
C15H16O2
133027
Bisphenol A, 97%
C15H16O2
54680
Butyl 4-hydroxybenzoate, ≥99.0% (GC)
C11H14O3
W220302
Butyl 4-hydroxybenzoate, ≥99%
C11H14O3
B1217000 Butyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
C11H14O3
1084000 Butylparaben, United States Pharmacopeia (USP) Reference Standard
C11H14O3
PHR1022 Butylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
C11H14O3
36141
Linuron, PESTANAL®, analytical standard
C9H10Cl2N2O2
P2155001
Plastic additive 01, European Pharmacopoeia (EP) Reference Standard
C24H38O4
64947
Prochloraz, certified reference material, TraceCERT®
C15H16Cl3N3O2
45631
Prochloraz, PESTANAL®, analytical standard
C15H16Cl3N3O2
45705
Vinclozolin, PESTANAL®, analytical standard
C12H9Cl2NO3