EMAIL THIS PAGE TO A FRIEND
Journal of gastroenterology and hepatology
Pretransplant replacement of donor liver grafts with recipient Kupffer cells attenuates liver graft rejection in rats.
PMID 25532540
Abstract
Rejection of liver grafts is a difficult issue that has not been resolved. Preoperative replacement of liver cells in the graft with cells from the intended recipient may attenuate rejection. We investigated whether preoperative transplant of recipient bone marrow cells (BMCs) to the donor replaced liver allograft cells and attenuated rejection. We used a rat model of allogeneic liver transplant (LT) from Dark Agouti (DA) to Lewis (LEW) rats. In BMC group, DA rats received BMC transplants from LacZ-transgenic LEW rats at 1 week before LT. In the control group, DA rats received no preoperative treatment. We evaluated graft damage at 7 days after LT and the survival of the recipient rats. Rats in the BMC group experienced prolonged survival that was abrogated by the administration of gadolinium chloride to donors at 24 h before LT. Serum concentrations of total bilirubin and hyaluronic acid on day 7 were significantly lower in the BMC group, and histopathological analyses revealed that rejection of the liver graft was attenuated. X-gal staining and immunohistostaining of the liver graft revealed that BMCs engrafted in the sinusoidal space differentiated into Kupffer cells. Preoperative transplant of recipient BMCs to LT donors replaced donor Kupffer cells and attenuated post-LT rejection, indicating that this strategy may increase the success of LT.
Related Materials