EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Development and co-validation of porcine insulin certified reference material by high-performance liquid chromatography-isotope dilution mass spectrometry.
PMID 25534116
Abstract
This article concerns the development and co-validation of a porcine insulin (pINS) certified reference material (CRM) produced by the National Institute of Metrology, People's Republic of China. Each CRM unit contained about 15xa0mg of purified solid pINS. The moisture content, amount of ignition residue, molecular mass, and purity of the pINS were measured. Both high-performance liquid chromatography-isotope dilution mass spectrometry and a purity deduction method were used to determine the mass fraction of the pINS. Fifteen units were selected to study the between-bottle homogeneity, and no inhomogeneity was observed. A stability study concluded that the CRM was stable for at least 12xa0months at -20xa0°C. The certified value of the CRM was (0.892 ± 0.036) g/g. A co-validation of the CRM was performed among Chinese, Japanese, and Korean laboratories under the framework of the Asian Collaboration on Reference Materials. The co-validation results agreed well with the certified value of the CRM. Consequently, the pINS CRM may be used as a calibration material or as a validation standard for pharmaceutical purposes to improve the quality of pharmaceutical products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
342041
Perfluoroheptanoic acid, 99%
C7HF13O2