EMAIL THIS PAGE TO A FRIEND
Nature reviews. Microbiology
Antimicrobials: New tricks for old drugs.
PMID 25534806
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33447
Mecillinam, VETRANAL, analytical standard
C15H23N3O3S