EMAIL THIS PAGE TO A FRIEND
Developmental neurobiology
Melanopsin ganglion cells extend dendrites into the outer retina during early postnatal development.
PMID 25534911
Abstract
Melanopsin ganglion cells express the photopigment melanopsin and are the first functional photoreceptors to develop in the mammalian retina. They have been shown to play a variety of important roles in visual development and behavior in the early postnatal period (Johnson et al., 2010; Kirkby and Feller, 2013; Rao et al., 2013; Renna et al., 2011). Here, we probed the maturation of the dendritic arbors of melanopsin ganglion cells during this developmental period in mice. We found that some melanopsin ganglion cells (mainly the M1-subtype) transiently extend their dendrites not only into the inner plexiform layer (where they receive synaptic inputs from bipolar and amacrine cells) but also into the outer plexiform layer, where in mature retina, rod and cone photoreceptors are thought to contact only bipolar and horizontal cells. Thus, some immature melanopsin ganglion cells are biplexiform. This feature is much less common although still present in the mature retina. It reaches peak incidence 8-12 days after birth, before the eyes open and bipolar cells are sufficiently mature to link rods and cones to ganglion cells. At this age, some outer dendrites of melanopsin ganglion cells lie in close apposition to the axon terminals of cone photoreceptors and express a postsynaptic marker of glutamatergic transmission, postsynaptic density-95 protein (PSD-95). These findings raise the possibility of direct, monosynaptic connections between cones and melanopsin ganglion cells in the early postnatal retina. We provide a detailed description of the developmental profile of these processes and consider their possible functional and evolutionary significance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA001198
Anti-BTK antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA002028
Anti-BTK antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500372
Anti-BTK antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4502936
Anti-BTK antibody produced in rabbit, affinity isolated antibody
B0811
Anti-BTK, C-Terminal antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1400106
Anti-GATM antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB2701647
Anti-gatm antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA026077
Anti-GATM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103200
Anti-GATM antibody produced in rabbit, affinity isolated antibody
SAB4503405
Anti-HAT, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1406820
Anti-TMPRSS11D antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV46418
Anti-TMPRSS11D antibody produced in rabbit, IgG fraction of antiserum
SAB2102485
Anti-TMPRSS11D antibody produced in rabbit, affinity isolated antibody
HPA052834
Anti-TMPRSS11D antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB5300137
Monoclonal Anti-BTK antibody produced in mouse, clone 7F12H4, ascites fluid
SAB1402784
Monoclonal Anti-GATM, (N-terminal) antibody produced in mouse, clone 2H7, ascites fluid