EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
The liver-selective NO donor, V-PYRRO/NO, protects against liver steatosis and improves postprandial glucose tolerance in mice fed high fat diet.
PMID 25534988
Abstract
There is an unmet medical need for novel NAFLD treatments. Here we have examined the effects of liver-selective NO donor (V-PYRRO/NO) as compared with metformin on hepatic steatosis and glucose tolerance in mice fed high fat diet. Effects of V-PYRRO/NO (5 mgkg(-1)) or metformin (616 mgkg(-1)) were examined in C57BL/6J mice fed high fat diet (HF, 60 kcal% fat). Quantitative determination of steatosis, liver fatty acid composition and western blot analysis of selected proteins involved in mitochondrial biogenesis, fatty acid de novo synthesis and oxidation, triacylglycerols and cholesterol transport from the liver were performed. Liver NOx and nitrate concentration and blood biochemistry were also analyzed. V-PYRRO/NO and metformin reduced liver steatosis with simultaneous reduction of total liver triacylglycerols, diacylglycerols and ceramides fraction and reversed HF-induced decrease in UFA/SFA ratio. V-PYRRO/NO substantially improved postprandial glucose tolerance, while the effect of metformin was modest and more pronounced on HOMA IR index. The anti-steatotic mechanism of V-PYRRO/NO was dependent on NO release, differed from that of metformin and involved improved glucose tolerance and inhibition of de novo fatty acid synthesis by Akt activation and ACC phosphorylation. In turn, major mechanism of metformin action involved increased expression of proteins implicated in mitochondrial biogenesis and metabolism (PGC-1α, PPARα, COX IV, cytochrome c, HADHSC). V-PYRRO/NO acts as a liver-specific NO donor prodrug affording pronounced anti-steatotic effects and may represent an efficient, mechanistically novel approach to prevent liver steatosis and insulin resistance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
UC126
4-Hydroxymephenytoin
C12H14N2O3
H146
4-Hydroxymephenytoin, ≥98% (HPLC)
C12H14N2O3
158968
α-D-Glucose, anhydrous, 96%
C6H12O6
G8769
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
C6H12O6
G8644
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
C6H12O6
G3285
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
C6H12O6
49163
Glucose solution, BioUltra, for molecular biology, ~20% in H2O
C6H12O6
357162
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
V
357170
Vanadium, foil, thickness 0.25 mm, 99.7% trace metals basis
V
266205
Vanadium, foil, thickness 0.5 mm, 99.7% trace metals basis
V
262935
Vanadium, powder, −325 mesh, 99.5% trace metals basis
V
266175
Vanadium, rod, diam. 6.2 mm, 99.7% trace metals basis
V
262927
Vanadium, turnings, 99.7% trace metals basis
V
774073
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
V
774065
Vanadium, powder, -100 mesh, 99.9% trace metals basis
V
GF98785388
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF25800386
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
V
GF46548692
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF21697106
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF76957004
Vanadium, microfoil, disks, 25mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF23355996
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF69763977
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
V
GF75438445
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%
V
GF84027269
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
V
GF25047503
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
V
GF39723239
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
V
GF29805341
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%
V
GF03408555
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
V
GF30937732
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
V
GF57656391
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
V