EMAIL THIS PAGE TO A FRIEND
Anti-cancer drugs
Chemotherapeutic effect of tamoxifen on temozolomide-resistant gliomas.
PMID 25535979
Abstract
Tamoxifen, a selective estrogen receptor modulator, is widely used in the chemotherapy of estrogen receptor-positive breast cancer. Recent studies have indicated that tamoxifen might have a potential chemotherapeutic effect on glioma. In the present study, we determined the chemotherapeutic action of tamoxifen on human glioma cell lines. Methylation of 06-methylguanine-DNA methyltransferase was identified in A172, U251, and BT325 glioma cell lines, but not in the U87 cell line. Consistently, A172, U251, and BT325 cell lines are resistant to temozolomide. Tamoxifen induced significant cytotoxic action in A172, U251, BT325, and U87 cell lines. Further, Hoechst 33342 staining and apoptosis flow cytometric analysis demonstrated that tamoxifen induced apoptosis in the BT325 cell line. Mitochondrial complex analysis indicated that tamoxifen, but not other estrogen receptor modulators, dose-dependently inhibits complex I activity. In summary, our study suggests that tamoxifen might have a chemotherapeutic effect on temozolomide-resistant glioma through its direct action on mitochondrial complex I inhibition and could provide further evidence to support future clinical trials of tamoxifen for the treatment of glioblastoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H6036 1,3,5-Tris(4-hydroxyphenyl)-4-propyl-1H-pyrazole, powder, ≥98% (HPLC)
C24H22N2O3