EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Microcytic anemia.
PMID 25539122
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3518
Carbonyl iron, ≥97% Fe basis
Fe
44890
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Fe
12310
Iron, ≥99%, reduced, powder (fine)
Fe
267945
Iron, chips, 99.98% trace metals basis
Fe
338141
Iron, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Fe
356808
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Fe
413054
Iron, granular, 10-40 mesh, >99.99% trace metals basis
Fe
513423
Iron, activated, nanoparticles, dispersion, <100 nm particle size, in mineral oil, 99.5% trace metals basis
Fe
IRMM524A
Iron, IRMM® certified Reference Material, 0.1 mm foil
Fe
IRMM524B
Iron, IRMM® certified Reference Material, 0.5 mm wire
Fe
255637
Iron, powder, ≥99.99% trace metals basis
Fe
209309
Iron, powder, −325 mesh, 97%
Fe
267953
Iron, powder, <10 μm, ≥99.9% trace metals basis
Fe
266213
Iron, rod, diam. 6.3 mm, 99.98% trace metals basis
Fe
266256
Iron, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Fe
266248
Iron, wire, diam. 0.5 mm, ≥99.99%
Fe
356824
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Fe
356832
Iron, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Fe
GF58945078
Iron, wire reel, 250m, diameter 0.25mm, hard, 99.5%
Fe
GF21695722
Iron, wire reel, 2m, diameter 0.13mm, hard, 99.99+%
Fe
GF81282135
Iron, wire reel, 2m, diameter 0.25mm, annealed, 99.99+%
Fe
GF57565996
Iron, wire reel, 2m, diameter 0.25mm, hard, 99.99+%
Fe
GF48022152
Iron, wire reel, 2m, diameter 0.5mm, hard, 99.99+%
Fe
GF48020947
Iron, wire reel, 500m, diameter 0.125mm, hard, 99.5%
Fe
GF58809965
Iron, wire reel, 50m, diameter 0.065mm, hard, 99.5%
Fe
GF34071566
Iron, wire reel, 50m, diameter 0.25mm, hard, 99.5%
Fe
GF96915134
Iron, wire reel, 50m, diameter 0.5mm, as drawn, 99.95%
Fe
GF35663935
Iron, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Fe
GF27511781
Iron, wire reel, 5m, diameter 0.25mm, annealed, 99.99+%
Fe
GF61040023
Iron, wire reel, 5m, diameter 0.25mm, hard, 99.99+%
Fe