EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Selective binding to monoamine oxidase A: in vitro and in vivo evaluation of (18)F-labeled β-carboline derivatives.
PMID 25541203
Abstract
In this study we synthesized four different (18)F-labeling precursors for the visualization of the monoamino oxidase A using harmol derivatives. Whereas two are for prosthetic group labeling using [(18)F]fluoro-d2-methyl tosylate and 2-[(18)F]fluoroethyl-tosylate, the other three precursors are for direct nucleophilic (18)F-labeling. Additionally the corresponding reference compounds were synthesized. The syntheses of [(18)F]fluoro-d2-methyl-harmol and 2-[(18)F]fluoroethyl-harmol were carried out using harmol as starting material. For direct nucleophilic (18)F-labeling of the tracers carrying oligoethyled spacers (PEG), a toluenesulfonyl leaving group was employed. The radiolabeling, purification and formulation for each tracer was optimized and evaluated in vitro and in vivo. Stability tests in human serum showed that all tracers were stable over the observation period of 60 min. μPET studies using of the synthesized tracers revealed that the tracers carrying PEG spacers showed no sufficient brain uptake. Consequently, the (18)F-fuoro alkylated tracers [(18)F]fluoro-d2-methyl-harmol and 2-[(18)F]fluoroethyl-harmol were further evaluated showing SUVs in the brain of 1.0±0.2 g/mL and 3.4±0.5 g/mL after 45 min, respectively. In blockade studies the selectivity and specificity of both tracers were demonstrated. However, for [(18)F]fluoro-d2-methyl-harmol a rapid washout from the brain was also observed. In vitro binding assays revealed that 2-[(18)F]fluoroethyl-harmol (IC50=0.54±0.06 nM) has a higher affinity than the (18)F-fluoro-d2-methylated ligand (IC50=12.2±0.6 nM), making 2-[(18)F]fluoroethyl-harmol superior to the other evaluated compounds and a promising tracer for PET imaging of the MAO A.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
746371
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
INa