EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Perovskite for the highly selective enrichment of phosphopeptides.
PMID 25542702
Abstract
Selective and effective enrichment of phosphopeptides from complex samples is essential in phosphoproteome study by mass spectrometry (MS). In this work, we compared perovskites (MgTiO3, CaTiO3, SrTiO3, BaTiO3 and CaZrO3) with metal oxides (ZrO2 and TiO2) in their capability for the selective enrichment of phosphopeptides. It was found here that perovskites exhibited higher selectivity towards phosphopeptides than commonly used ZrO2 and TiO2, even though they all have high affinity to phosphopeptides. As for perovskites, CaTiO3 exhibited better selectivity for enrichment of phosphopeptides than SrTiO3, MgTiO3, BaTiO3 and CaZrO3, which might be ascribed to their crystal structures and electrophilic abilities. Moreover, to further confirm the performance of CaTiO3, CaTiO3 and TiO2 were applied to the enrichment of phosphopeptides from tryptic digest of proteins of human Jurkat-T cell lysate, respectively. The results showed CaTiO3 has much higher selectivity than TiO2 in the enrichment of phosphopeptides from the complex biological sample. Taken together, here we show that CaTiO3 is an excellent material for the highly selective enrichment of phosphopeptides and it could be potentially used in the large-scale phosphoproteome study.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
39319
2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
C7H6O4
78069
2,5-Dihydroxybenzoic acid, analytical standard
C7H6O4
45533
3-Indoleacetic acid, PESTANAL®, analytical standard
C10H9NO2
677469
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
O2Ti
718467
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
O2Ti
774510
Titanium(IV) oxide, nanowires, diam. × L ~100 nm × 10 μm
O2Ti
774529
Titanium(IV) oxide, nanowires, diam. × L ~10 nm × 10 μm
O2Ti
637254
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
O2Ti
232033
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
O2Ti
248576
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
O2Ti
634662
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
O2Ti
700355
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticle, <250 nm particle size (DLS), paste, 53-57 wt. % in diethylene glycol monobutyl ether/ethylene glycol, 99.9% trace metals basis
O2Ti
700347
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 33-37 wt. % in H2O, 99.5% trace metals basis
O2Ti
700339
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <100 nm particle size, dispersion, 48-52 wt. % in xylene, 99.9% trace metals basis
O2Ti
204730
Titanium(IV) oxide, rutile, 99.995% trace metals basis
O2Ti
204757
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
O2Ti
635057
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), 99.99% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
O2Ti
635049
Titanium(IV) oxide, rutile, <100>, (single crystal substrate), 99.99% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
O2Ti
635065
Titanium(IV) oxide, rutile, <110>, single crystal substrate, 99.99% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
O2Ti
637262
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
O2Ti
224227
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
O2Ti