EMAIL THIS PAGE TO A FRIEND
Cancer research
HOXB7 promotes malignant progression by activating the TGFβ signaling pathway.
PMID 25542862
Abstract
Overexpression of HOXB7 in breast cancer cells induces an epithelial-mesenchymal transition and promotes tumor progression and lung metastasis. However, the underlying mechanisms for HOXB7-induced aggressive phenotypes in breast cancer remain largely unknown. Here, we report that phosphorylation of SMAD3 was detected in a higher percentage in primary mammary tumor tissues from double-transgenic MMTV-Hoxb7/Her2 mice than tumors from single-transgenic Her2/neu mice, suggesting activation of TGFβ/SMAD3 signaling by HOXB7 in breast tumor tissues. As predicted, TGFβ2 was high in four MMTV-Hoxb7/Her2 transgenic mouse tumor cell lines and two breast cancer cell lines transfected with HOXB7, whereas TGFβ2 was low in HOXB7-depleted cells. HOXB7 directly bound to and activated the TGFβ2 promoter in luciferase and chromatin immunoprecipitation assays. Increased migration and invasion as a result of HOXB7 overexpression in breast cancer cells were reversed by knockdown of TGFβ2 or pharmacologic inhibition of TGFβ signaling. Furthermore, knockdown of TGFβ2 in HOXB7-overexpressing MDA-MB-231 breast cancer cells dramatically inhibited metastasis to the lung. Interestingly, HOXB7 overexpression also induced tumor-associated macrophage (TAM) recruitment and acquisition of an M2 tumor-promoting phenotype. TGFβ2 mediated HOXB7-induced activation of macrophages, suggesting that TAMs may contribute to HOXB7-promoted tumor metastasis. Providing clinical relevance to these findings, by real-time PCR analysis, there was a strong correlation between HOXB7 and TGFβ2 expression in primary breast carcinomas. Taken together, our results suggest that HOXB7 promotes tumor progression in a cell-autonomous and non-cell-autonomous manner through activation of the TGFβ signaling pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN10247 CompoZr® Knockout ZFN Kit, Human HOXB7 (NM_004502)