EMAIL THIS PAGE TO A FRIEND
Journal of wound care
Measurement of microelectric potentials in a bioelectrically-active wound care device in the presence of bacteria.
PMID 25543820
Abstract
Wound healing is enhanced in the presence of an external electrical field. The purpose of this study was first to investigate whether microelectric potentials (EPs) can be generated when the innovative design of a silver (Ag)- and zinc (Zn)-printed wound care device was exposed to saline solution which is commonly used to clean wound sites; and second to measure the generated EPs while the device was exposed to bacterial culture suspensions to mimic infection. An Ag/Zn-printed test specimen as a wound care device was designed to accommodate Ag and Zn half-cell potentials by alternatively printing them on a woven polyester material in a well-characterised dot matrix pattern. A well-calibrated high impedance EPs measuring system was used to measure any EPs generated. Ultrasensitive inductively coupled plasma analysis was performed to determine whether the device induced any increase in trace metals in rabbit blood following implantation for 2-4 weeks. EPs were consistently generated under various conductive solutions at the levels of 120.4±26.3mV (average±standard deviation) on Ag dots and -506.5±76.3mV over Zn dots to form microcircuits with EPs of 626.7±86.3mV between the Ag and Zn metallic elements of the dressing. Interestingly, the patterns of EPs generated with stable polarities were consistent when the device was exposed to bacterial suspensions for mimicking wound infection. Implantation of the device did not cause any increase in Ag or Zn in rabbit blood. The Ag/Zn-printed wound device generated sustained EPs successfully in the presence of various conductive fluids without changing EPs including polarities. Consistently generated EPs at each battery couple with Ag/Zn-based wound device would restore disrupted physiologic bioelectric signals on wound sites, which could lead to improved wound healing. The opinions or assertions contained herein are the private views of the authors and are not to be construed as official or as reflecting the views of the US Department of the Army, the US Department of Defense, the Department of Veterans Affairs, or the Henry M. Jackson Foundation for Advancement of Military Medicine. The research was supported by the US Air Force medical support agency (AFMSA), office of the Air Force Surgeon General, USA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
85131
Silver, colloidal, 65-75% Ag basis
Ag
373249
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Ag
327077
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Ag
326976
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Ag
348724
Silver, foil, thickness 1.5 mm, 99.9% trace metals basis
Ag
369438
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Ag
265543
Silver, foil, thickness 1.0 mm, 99.9% trace metals basis
Ag
265535
Silver, foil, thickness 0.5 mm, 99.99% trace metals basis
Ag
345075
Silver, foil, thickness 0.5 mm, 99.9% trace metals basis
Ag
348716
Silver, foil, thickness 0.25 mm, 99.99% trace metals basis
Ag
326984
Silver, foil, thickness 0.25 mm, 99.9% trace metals basis
Ag
265527
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Ag
326992
Silver, foil, thickness 0.075 mm, 99.9% trace metals basis
Ag
265519
Silver, foil, thickness 0.025 mm, 99.9% trace metals basis
Ag
303372
Silver, granular, >250 μm, 99.99% trace metals basis
Ag
461032
Silver, needles, 250-600 μm, ≥99.99% trace metals basis
Ag
265500
Silver, powder, <250 μm, 99.99% trace metals basis
Ag
327107
Silver, powder, <45 μm, ≥99.99% trace metals basis
Ag
327093
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Ag
327085
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Ag
484059
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Ag
576832
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Ag
265624
Silver, rod, diam. 7.0 mm, 99.99% trace metals basis
Ag
348775
Silver, rod, diam. 3.2 mm, 99.98% trace metals basis
Ag
204366
Silver, shot, 1-3 mm, 99.999% trace metals basis
Ag
327050
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Ag
267430
Silver, wire, diam. 2.0 mm, ≥99.99% trace metals basis
Ag
327018
Silver, wire, diam. 2.0 mm, 99.9% trace metals basis
Ag
348759
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Ag
265616
Silver, wire, diam. 1.5 mm, 99.9% trace metals basis
Ag