EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Adsorption of water from aqueous acetonitrile on silica-based stationary phases in aqueous normal-phase liquid chromatography.
PMID 25544246
Abstract
Excess adsorption of water from aqueous acetonitrile mobile phases was investigated on 16 stationary phases using the frontal analysis method and coulometric Karl-Fischer titration. The stationary phases include silica gel and silica-bonded phases with different polarities, octadecyl and cholesterol, phenyl, nitrile, pentafluorophenylpropyl, diol and zwitterionic sulfobetaine and phosphorylcholine ligands bonded on silica, hybrid organic-silica and hydrosilated matrices. Both fully porous and core-shell column types were included. Preferential uptake of water by the columns can be described by Langmuir isotherms. Even though a diffuse rather than a compact adsorbed discrete layer of water on the adsorbent surface can be formed because of the unlimited miscibility of water with acetonitrile, for convenience, the preferentially adsorbed water was expressed in terms of a hypothetical monomolecular water layer equivalent in the inner pores. The uptake of water strongly depends on the polarity and type of the column. Less than one monomolecular water layer equivalent was adsorbed on moderate polar silica hydride-based stationary phases, Ascentis Express F5 and Ascentis Express CN column at the saturation capacity, while on more polar stationary phases, several water layer equivalents were up-taken from the mobile phase. The strongest affinity to water was observed on the ZIC cHILIC stationary phases, where more than nine water layer equivalents were adsorbed onto its surface at its saturation capacity. Columns with bonded hydroxyl and diol ligands show stronger water adsorption in comparison to bare silica. Columns based on hydrosilated silica generally show significantly decreased water uptake in comparison to stationary phases bonded on ordinary silica. Significant correlations were found between the water uptake and the separation selectivity for compounds with strong polarity differences.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09689
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
C2H7NO2
73594
Ammonium acetate, for mass spectrometry, eluent additive for LC-MS
C2H7NO2
09691
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
C2H7NO2
536164
Ethylene, 99.99%
C2H4
00489
Ethylene, puriss., ≥99.95% (GC)
C2H4
03482
Ethylene, purum, ≥99.9%
C2H4
03484
Ethylene, ≥99.9%
C2H4
295329
Ethylene, ≥99.5%
C2H4
Y0000764
Fluorouracil impurity C, European Pharmacopoeia (EP) Reference Standard
C4H4N2O2
PHR1317
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
C7H8
87362
Tetrahydrofuran, for luminescence, ≥99.5% (GC)
C4H8O
87363
Tetrahydrofuran, for amino acid analysis, without stabilizer
C4H8O
87369
Tetrahydrofuran, Selectophore, ≥99.5%
C4H8O
78445
Tetrahydrofuran, analytical standard
C4H8O
33709
Tetrahydrofuran, puriss. p.a., ≥99.9% (GC)
C4H8O
87370
Tetrahydrofuran, purum, ≥99.0% (GC)
C4H8O
PHR1561
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
C4H8O
T7875
Thiourea, ReagentPlus®, ≥99.0%
CH4N2S
T8656
Thiourea, ACS reagent, ≥99.0%
CH4N2S
88810
Thiourea, puriss. p.a., ACS reagent, ≥99.0%
CH4N2S
89680
Toluene, analytical standard
C7H8
PHR1311
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
C7H8
U0750
Uracil, ≥99.0%
C4H4N2O2
U1128
Uracil, suitable for cell culture, BioReagent
C4H4N2O2
94220
Uracil, ≥99.0% (T)
C4H4N2O2
PHR1581
Uracil, Pharmaceutical Secondary Standard; Certified Reference Material
C4H4N2O2