EMAIL THIS PAGE TO A FRIEND
Tuberculosis (Edinburgh, Scotland)
Arabinosylated lipoarabinomannan (Ara-LAM) mediated intracellular mechanisms against tuberculosis infection: involvement of protein kinase C (PKC) mediated signaling.
PMID 25544312
Abstract
Tuberculosis causes severe immunosuppression thereby ensuring the loss of the host protective immune responses. During Mycobacterium tuberculosis infection, the pathogen modulates TLR-2 receptor down-stream signaling, indicating the possible involvement of TLR-2 in the regulation of the host immune response. Moreover, different PKC isoforms are also involved in the course of infection. Arabinosylated lipoarabinomannan (Ara-LAM) possesses immuno-modulatory properties which induce the pro-inflammatory responses via induction of TLR-2-mediated signaling. Here, we found that pretreatment of M.xa0tuberculosis-infected macrophages with Ara-LAM caused a significant increase in the conventional PKC expression along with their active association with TLR-2. This association activated the TLR-2 -mediated downstream signaling, facilitating the activation of MAP kinase P38. All these events culminated in the up-regulation of proinflammatory response, which was abrogated by treatment with PKC-α and P38 inhibitors. Moreover, pretreatment of macrophages with Ara-LAM abrogated the IL-10 production while restored MHC-II expression in the infected macrophages. This study demonstrates that Ara-LAM confers protection against tuberculosis via TLR-2/PKC signaling crosstalk which is responsible for the induction of host protective immune response against tuberculosis.