PloS one
Cdk1-mediated phosphorylation of human ATF7 at Thr-51 and Thr-53 promotes cell-cycle progression into M phase.
PMID 25545367
Abstract
Activating transcription factor 2 (ATF2) and its homolog ATF7 are phosphorylated at Thr-69/Thr-71 and at Thr-51/Thr-53, respectively, by stress-activated MAPKs regulating their transcriptional functions in G1 and S phases. However, little is known about the role of ATF2 and ATF7 in G2/M phase. Here, we show that Cdk1-cyclin B1 phosphorylates ATF2 at Thr-69/Thr-71 and ATF7 at Thr-51/Thr-53 from early prophase to anaphase in the absence of any stress stimulation. Knockdown of ATF2 or ATF7 decreases the rate of cell proliferation and the number of cells in M-phase. In particular, the knockdown of ATF7 severely inhibits cell proliferation and G2/M progression. The inducible expression of a mitotically nonphosphorylatable version of ATF7 inhibits G2/M progression despite the presence of endogenous ATF7. We also show that mitotic phosphorylation of ATF7 promotes the activation of Aurora kinases, which are key enzymes for early mitotic events. These results suggest that the Cdk1-mediated phosphorylation of ATF7 facilitates G2/M progression, at least in part, by enabling Aurora signaling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
40311
2-Butanone, 5000 μg/mL in methanol: water (9:1), analytical standard
C4H8O
02469
2-Butanone, analytical standard
C4H8O
W217018
2-Butanone, ≥99.5%, FCC, FG
C4H8O
48877
2-Butanone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
C4H8O
32883
4-tert-Octylphenol monoethoxylate solution, 1 μg/mL in acetone, analytical standard
C16H26O2
32882
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
C16H26O2
MAB1501
Anti-Actin Antibody, clone C4, 100 µL, Reliably and specifically detect actin using this Anti-Actin Antibody, clone C4. This highly published monoclonal antibody is validated for use in ELISA, IC, IF, IH, IH(P) & WB. This mAb is also available as a fluorescent conjugate.
A4086 Anti-ATF-2 (SS-16) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2500131
Anti-ATF7 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA003384
Anti-ATF7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
47072
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
FNa
80373
Fluoride, certified anion standard solution, 1000 mg/kg F-, for ion chromatography, certified by BAM, according to ISO 17025
FNa
1295607
Glycerin, United States Pharmacopeia (USP) Reference Standard
C3H8O3
PHR1020
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O3
49779
Glycerol, tested according to Ph.Eur., anhydrous
C3H8O3
77067
Glycerol, analytical standard
C3H8O3
12146
Glycerol 3 M solution, 3 M
C3H8O3 · xH2O
54461
HEPES, ≥99.0% (T)
C8H18N2O4S
PHR1428
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
C8H18N2O4S
PHR1207
Methyl Ethyl Ketone, Pharmaceutical Secondary Standard; Certified Reference Material
C4H8O
C4710
Monoclonal Anti-Cyclin A antibody produced in mouse, clone CY-A1, ascites fluid
450022
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
FNa
PHR1408
Sodium Fluoride, certified reference material
FNa
67414
Sodium fluoride 0.5 M solution
FNa
T1895
Thymidine, powder, BioReagent, suitable for cell culture
C10H14N2O5
T9250
Thymidine, ≥99%
C10H14N2O5
89270
Thymidine, ≥99.0% (HPLC)
C10H14N2O5