EMAIL THIS PAGE TO A FRIEND
Inflammatory bowel diseases
Validation of gene expression biomarker analysis for biopsy-based clinical trials in Crohn's disease.
PMID 25545378
Abstract
The ability to measure the expression of proinflammatory cytokines from intestinal biopsies in patients with Crohn's disease in an accurate and reproducible way is critical for proof-of-concept and mechanism-of-action trials; however, the number of biopsies from a segment of the ileum or colon required to yield reproducible results has not been rigorously evaluated. We examined intestinal biopsies from patients with Crohn's disease to validate methods for detecting changes in inflammatory gene expression. To evaluate the reproducibility of gene expression measurements, intestinal biopsies were obtained from designated segments from 6 healthy controls, 6 patients with active Crohn's disease, and 6 patients with inactive Crohn's disease. Disease activity was based on the simple endoscopic score for Crohn's disease. Expression of 7 proinflammatory genes was measured from each biopsy using quantitative polymerase chain reaction. Using a linear mixed effects model, the power to detect transcriptional changes corresponding to active and inactive Crohn's disease was calculated. Total simple endoscopic score for Crohn's disease score corresponds with expression of most inflammatory biomarkers. For most genes, 2 to 5 biopsies are needed to reduce sampling error to <25% for most genes. To measure changes in mRNA expression corresponding to active versus inactive Crohn's disease, 1 to 2 intestinal biopsies from 3 patients before and after treatment are needed to yield power of at least 80%. Measuring proinflammatory gene expression from mucosal biopsies from patients with Crohn's disease is practicable and provides objective biomarkers that can be used in proof-of-concept and mechanism-of-action trials to assess response to therapy.