EMAIL THIS PAGE TO A FRIEND
Journal of cellular physiology
Hepatic regenerative potential of mouse bone marrow very small embryonic-like stem cells.
PMID 25545634
Abstract
Very small embryonic-like stem cells (VSELs) are a Sca-1 (+)  Lin(-) CD45(-) cell population that has been isolated from the bone marrow of mice. The similarities and differences between the mRNA profiles of VSELs and embryonic stem (ES) cells have not yet been defined. Here, we report the whole genome gene expression profile of VSELs and ES cells. We analyzed the global gene expression of VSELs and compared it with ES cells by microarray analysis. We observed that 9,521 genes are expressed in both VSELs and ES cells, 1,159 genes are expressed uniquely in VSELs, and 420 genes are expressed uniquely in ES cells. We found that although VSELs are similar to ES cells in their expression of genes associated with stem cell behavior and pluripotency, there are also differences in their mRNA expression. We further analyzed the expression of stem cell-associated genes in VSELs and ES cells, and found that there were differences in these genes. For instance, the Pkd2 and Yap1 gene were reduced in their expression in VSELs when compared with ES cells. But we also found Zfp54 gene expression was higher in VSELs compared with ES cells. More interestingly, we demonstrated that VSELs express c-kit, the stem cell factor (SCF) receptor. In vitro, SCF promoted VSEL differentiation into hepatic colonies in the presence of hepatocyte growth factor. In vivo, transplantation of VSELs directly into CCl4-induced injured livers significantly reduced serum ALT and AST levels. Therefore, these data suggest that VSELs play a role in the repair of injured livers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300489
Anti-KIT (Ab-721) antibody produced in rabbit, affinity isolated antibody
SAB4300593
Anti-KIT (Ab-936) antibody produced in rabbit, affinity isolated antibody
GW21114 Anti-KIT antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1412042
ANTI-KIT antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
SAB1412043
ANTI-KIT antibody produced in mouse, clone 5F6, purified immunoglobulin, buffered aqueous solution
SAB1412044
ANTI-KIT antibody produced in mouse, clone 5A11, purified immunoglobulin, buffered aqueous solution
SAB1412045
ANTI-KIT antibody produced in mouse, clone 2C3, purified immunoglobulin, buffered aqueous solution
SAB1412046
ANTI-KIT antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
SAB1412140
ANTI-KIT antibody produced in mouse, clone 4F7, purified immunoglobulin, buffered aqueous solution
SAB1412141
ANTI-KIT antibody produced in mouse, clone 3A8, purified immunoglobulin, buffered aqueous solution
HPA004471
Anti-KIT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501647
Anti-KIT, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4700750
Monoclonal Anti-CD117 antibody produced in mouse, clone 104D2, purified immunoglobulin, buffered aqueous solution
WH0003815M2
Monoclonal Anti-KIT antibody produced in mouse, clone 6F2, purified immunoglobulin, buffered aqueous solution
AMAB90901
Monoclonal Anti-KIT antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1657, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90904
Monoclonal Anti-KIT antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1667, purified immunoglobulin, buffered aqueous glycerol solution
SAB1404007
Monoclonal Anti-KIT antibody produced in mouse, clone 6G12, purified immunoglobulin, buffered aqueous solution
SAB5300439
Monoclonal Anti-KIT antibody produced in mouse, clone 8D7, ascites fluid
SAB5300440
Monoclonal Anti-KIT antibody produced in mouse, clone 1C5, ascites fluid