EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Human mutations in methylenetetrahydrofolate dehydrogenase 1 impair nuclear de novo thymidylate biosynthesis.
PMID 25548164
Abstract
An inborn error of metabolism associated with mutations in the human methylenetetrahydrofolate dehydrogenase 1 (MTHFD1) gene has been identified. The proband presented with SCID, megaloblastic anemia, and neurologic abnormalities, but the causal metabolic impairment is unknown. SCID has been associated with impaired purine nucleotide metabolism, whereas megaloblastic anemia has been associated with impaired de novo thymidylate (dTMP) biosynthesis. MTHFD1 functions to condense formate with tetrahydrofolate and serves as the primary entry point of single carbons into folate-dependent one-carbon metabolism in the cytosol. In this study, we examined the impact of MTHFD1 loss of function on folate-dependent purine, dTMP, and methionine biosynthesis in fibroblasts from the proband with MTHFD1 deficiency. The flux of formate incorporation into methionine and dTMP was decreased by 90% and 50%, respectively, whereas formate flux through de novo purine biosynthesis was unaffected. Patient fibroblasts exhibited enriched MTHFD1 in the nucleus, elevated uracil in DNA, lower rates of de novo dTMP synthesis, and increased salvage pathway dTMP biosynthesis relative to control fibroblasts. These results provide evidence that impaired nuclear de novo dTMP biosynthesis can lead to both megaloblastic anemia and SCID in MTHFD1 deficiency.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA015006
Anti-MTHFD1 antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
N5755
β-Nicotinamide adenine dinucleotide phosphate hydrate
C21H28N7O17P3 · xH2O
N8035
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 5 mg (per vial)
C21H27N7NaO17P3
N8160
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 10 mg (per vial)
C21H27N7NaO17P3
Y0000441
Didanosine impurity A,, European Pharmacopoeia (EP) Reference Standard
C5H4N4O
M9500
DL-Methionine, ≥99%
C5H11NO2S
M2768
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
C5H11NO2S
64340
DL-Methionine, ≥99.0% (NT)
C5H11NO2S
W330108
DL-Methionine, 99%, FCC, FG
C5H11NO2S
M0950000
DL-Methionine, European Pharmacopoeia (EP) Reference Standard
C5H11NO2S
Y0000764
Fluorouracil impurity C, European Pharmacopoeia (EP) Reference Standard
C4H4N2O2
77662
Hypoxanthine, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
C5H4N4O
H9377
Hypoxanthine, ≥99.0%
C5H4N4O
H9636
Hypoxanthine, powder, BioReagent, suitable for cell culture
C5H4N4O
M0960000
Methionine, European Pharmacopoeia (EP) Reference Standard
C5H11NO2S
82580
Purine, puriss., ≥98.5% (HPLC)
C5H4N4
P55805
Purine, 98%
C5H4N4
U0750
Uracil, ≥99.0%
C4H4N2O2
U1128
Uracil, suitable for cell culture, BioReagent
C4H4N2O2
94220
Uracil, ≥99.0% (T)
C4H4N2O2
PHR1581
Uracil, Pharmaceutical Secondary Standard; Certified Reference Material
C4H4N2O2