EMAIL THIS PAGE TO A FRIEND
Toxicon : official journal of the International Society on Toxinology
Detection of marine microalgal biotoxins using bioassays based on functional expression of tunicate xenobiotic receptors in yeast.
PMID 25549942
Abstract
Marine microalgae can produce biotoxins that cause widespread poisoning in marine ecosystems and may also affect human health. While established microalgal biotoxins are detectable using chemical methods, a need remains for robust, inexpensive bioassays. Ligand-binding domains (LBDs) from a tunicate nuclear receptor, VDR/PXRα, which is orthologous to both the vertebrate pregnane X receptor (PXR) and the vitamin D receptor (VDR), can be activated by microalgal biotoxins when expressed in mammalian cell lines. Building on this observation, we developed a generic recombinant yeast bioassay platform that expresses chimeric proteins containing tunicate VDR/PXRα LBDs which mediate ligand-dependent transcription of a reporter gene (lacZ) encoding an easily assayed enzyme (β-galactosidase). Recombinant yeast strains expressing VDR/PXRα LBDs from two tunicate species, Ciona intestinalis and Botryllus schlosseri, were exposed to both synthetic and natural toxins. Structurally simple synthetic chemicals (n-butyl-p-aminobenzoate, carbamazepine, p-aminobenzoic acid, and bisphenol-A) generated EC50 values in the μM range, while more structurally complex marine biotoxins (okadaic acid, pectenotoxin-11, and portimine) activated the assays in the nM range. Given the large number of tunicate species, we propose that tunicate VDR/PXR LBDs may be used as 'sensor elements' in similar yeast-based high-throughput bioassays for detection of established microalgal biotoxins and uncharacterised marine bioactive compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
199524
Chlorophenol Red, indicator grade
C19H12Cl2O5S
59767
Chlorophenol Red-β-D-galactopyranoside, ≥90% (HPLC)
C25H22Cl2O10S
A5474
L-Aspartic acid, PharmaGrade, Ajinomoto, EP, JP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C4H7NO4
A9256
L-Aspartic acid, reagent grade, ≥98% (HPLC)
C4H7NO4
A8949
L-Aspartic acid, BioXtra, ≥99% (HPLC)
C4H7NO4
A7219
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
C4H7NO4
A4534
L-Aspartic acid, BioReagent, suitable for cell culture, suitable for insect cell culture
C4H7NO4
11189
L-Aspartic acid, BioUltra, ≥99.5% (T)
C4H7NO4
W365601
L-Aspartic acid, ≥98%, FG
C4H7NO4
G5667
L-Glutamic acid, PharmaGrade, Ajinomoto, EP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C5H9NO4
W328502
L-Glutamic acid, 99%, FCC
C5H9NO4
G8415
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
C5H9NO4
49449
L-Glutamic acid, BioUltra, ≥99.5% (NT)
C5H9NO4
G1251
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
C5H9NO4
H6034
L-Histidine, cell culture tested, meets EP, USP testing specifications, from non-animal source
C6H9N3O2
H8000
L-Histidine, ReagentPlus®, ≥99% (TLC)
C6H9N3O2
H3911
L-Histidine, PharmaGrade, Ajinomoto, EP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C6H9N3O2
53319
L-Histidine, BioUltra, ≥99.5% (NT)
C6H9N3O2
W527602
L-Isoleucine, 99%, FCC, FG
C6H13NO2
I5281
L-Isoleucine, PharmaGrade, Ajinomoto, EP, JP, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C6H13NO2
I2752
L-Isoleucine, reagent grade, ≥98% (HPLC)
C6H13NO2
I7403
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
C6H13NO2
58879
L-Isoleucine, BioUltra, ≥99.5% (NT)
C6H13NO2
I2877
L-Isoleucine: D-allo-isoleucine
C6H13NO2
L8912
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
C6H13NO2
L8000
L-Leucine, reagent grade, ≥98% (HPLC)
C6H13NO2
L6914
L-Leucine, PharmaGrade, Ajinomoto, EP, JP, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C6H13NO2
61819
L-Leucine, BioUltra, ≥99.5% (NT)
C6H13NO2
W329703
L-Leucine, 99%, FG
C6H13NO2
M9625
L-Methionine, reagent grade, ≥98% (HPLC)
C5H11NO2S