EMAIL THIS PAGE TO A FRIEND
Anticancer research
Prognostic markers for patient outcome following vaccination with multiple MHC Class I/II-restricted WT1 peptide-pulsed dendritic cells plus chemotherapy for pancreatic cancer.
PMID 25550602
Abstract
Treatment combining dendritic cells (DCs) pulsed with three types of major histocompatibility complex (MHC) class I and II (DC/WT1-I/II)-restricted Wilms' tumor 1 (WT1) peptides with chemotherapy may stabilize disease in pancreatic cancer patients. Laboratory data from seven patients with pancreatic cancer who underwent combined DC/WT1-I/II vaccination and chemotherapy were analyzed. The DC phenotypes and plasma cytokine profiles were analyzed via flow cytometry. The post-treatment neutrophil to lymphocyte (N/L) ratio was a treatment-related prognostic factor for better survival. Moreover, the mean fluorescence intensities (MFIs) of human leukocyte antigen (HLA)-DR and cluster of differentiation (CD)83 on DCs were significantly increased after chemoimmunotherapy. Interestingly, interleukin (IL)-6 level in plasma was significantly increased after chemoimmunotherapy in non-super-responders. An increased N/L ratio, as well as HLA-DR and CD83 MFI levels may be prognostic markers of longer survival in patients with advanced pancreatic cancer who undergo chemoimmunotherapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3897
2′-Deoxycytidine, ≥99% (HPLC)
C9H13N3O4