EMAIL THIS PAGE TO A FRIEND
BMC developmental biology
Denervation impairs regeneration of amputated zebrafish fins.
PMID 25551555
Abstract
Zebrafish are able to regenerate many of its tissues and organs after damage. In amphibians this process is regulated by nerve fibres present at the site of injury, which have been proposed to release factors into the amputated limbs/fins, promoting and sustaining the proliferation of blastemal cells. Although some candidate factors have been proposed to mediate the nerve dependency of regeneration, the molecular mechanisms involved in this process remain unclear. We have used zebrafish as a model system to address the role of nerve fibres in fin regeneration. We have developed a protocol for pectoral fin denervation followed by amputation and analysed the regenerative process under this experimental conditions. Upon denervation fins were able to close the wound and form a wound epidermis, but could not establish a functional apical epithelial cap, with a posterior failure of blastema formation and outgrowth, and the accumulation of several defects. The expression patterns of genes known to be key players during fin regeneration were altered upon denervation, suggesting that nerves can contribute to the regulation of the Fgf, Wnt and Shh pathways during zebrafish fin regeneration. Our results demonstrate that proper innervation of the zebrafish pectoral fin is essential for a successful regenerative process, and establish this organism as a useful model to understand the molecular and cellular mechanisms of nerve dependence, during vertebrate regeneration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L9756
(−)-Tetramisole hydrochloride, ≥99% (GC)
C11H12N2S · HCl
D27802
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
C6H12N2
09972
3-Nitrotetrazolium blue chloride, ≥75% (AT)
C40H30Cl2N10O6
06-570
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, 200 µg, Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker is a Rabbit Polyclonal Antibody for detection of Histone H3 phosphorylated at serine 10.This highly published Ab, also known as Anti-H3S10p, has been validated in ICC, IP & WB.
290734
Dabco® 33-LV
C6H12N2
D9564
DAPI, dilactate, ≥98% (HPLC)
C16H15N5 · 2C3H6O3
E10521
Ethyl 3-aminobenzoate methanesulfonate, 98%
C9H11NO2 · CH4SO3
A5040
Ethyl 3-aminobenzoate methanesulfonate salt, analytical standard
C9H11NO2 · CH4SO3
1295607
Glycerin, United States Pharmacopeia (USP) Reference Standard
C3H8O3
PHR1020
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O3
49779
Glycerol, tested according to Ph.Eur., anhydrous
C3H8O3
77067
Glycerol, analytical standard
C3H8O3
12146
Glycerol 3 M solution, 3 M
C3H8O3 · xH2O
11383213001 NBT, 4-Nitro blue tetrazolium chloride, solution
84010
NBT Vials, for in vitro diagnostic use
C40H30Cl2N10O6
N6876
Nitrotetrazolium Blue chloride, ~98% (TLC)
C40H30Cl2N10O6
N6639
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
C40H30Cl2N10O6
74032
Nitrotetrazolium Blue chloride, ≥98.0% (calc. based on dry substance, T)
C40H30Cl2N10O6
P6556 Proteinase K from Tritirachium album, lyophilized powder, ≥30 units/mg protein
T1512
Tetramisole hydrochloride, phosphatase inhibitor
C11H12N2S · HCl
T4375
Tetrazolium Blue Chloride, used in colorimetric determination of reducing compounds
C40H32Cl2N8O2
88190
Tetrazolium Blue Chloride, indicator for germination, for microbiology, ≥90% (T)
C40H32Cl2N8O2