PloS one
PGC TagSNP and its interaction with H. pylori and relation with gene expression in susceptibility to gastric carcinogenesis.
PMID 25551587
Abstract
Pepsinogen C (PGC) plays an important role in sustaining the cellular differentiation during the process of gastric carcinogenesis. This study aimed to assess the role of PGC tagSNPs and their interactions with Helicobacter pylori (H. pylori) in the development of gastric cancer and its precursor, atrophic gastritis. Four PGC tagSNPs (rs6941539, rs6912200, rs3789210 and rs6939861) were genotyped by Sequenom MassARRAY platform in a total of 2311 subjects consisting of 642 gastric cancer, 774 atrophic gastritis, and 895 healthy control subjects. The mRNA and protein expression levels of PGC in gastric tissues and in serum were respectively measured by quantitative reverse transcriptase-polymerase chain reaction (qRT-PCR), immunohistochemistry, and Eenzyme-linked immunoabsorbent assay (ELISA). We found associations between PGC rs3789210 CG/GG genotypes and reduced gastric cancer risk and between PGC rs6939861 A variant allele and increased risks of both gastric cancer and atrophic gastritis. As for the haplotypes of PGC rs6941539-rs6912200-rs3789210-rs6939861 loci, the TTCA and TTGG haplotypes were respectively associated with increased and reduced risks of both gastric cancer and atrophic gastritis; additionally, the CTCA haplotype was associated with increased atrophic gastritis risk. Very interestingly, rs6912200 CT/TT genotypes had a positive interaction with H. pylori, synergistically elevating the gastric cancer risk. Moreover, healthy subjects who carried rs6912200 CT, TT and CT/TT variant genotypes had lower histological and serum expression levels of PGC protein. Our findings highlight an important role of PGC rs3789210 and rs6939861 in altering susceptibility to atrophic gastritis and/or gastric cancer. Moreover, people who carry rs6912200 variant genotypes exhibit higher gastric cancer risk in case of getting H. pylori infection, which strongly suggest a necessity of preventing and/or eliminating H. pylori infection in those individuals.