EMAIL THIS PAGE TO A FRIEND
Human molecular genetics
Toll-like receptor 4 ablation in mdx mice reveals innate immunity as a therapeutic target in Duchenne muscular dystrophy.
PMID 25552658
Abstract
Toll-like receptor 4 (TLR4) recognizes specific structural motifs associated with microbial pathogens and also responds to certain endogenous host molecules associated with tissue damage. In Duchenne muscular dystrophy (DMD), inflammation plays an important role in determining the ultimate fate of dystrophic muscle fibers. In this study, we used TLR4-deficient dystrophic mdx mice to assess the role of TLR4 in the pathogenesis of DMD. TLR4 expression was increased and showed enhanced activation following agonist stimulation in mdx diaphragm muscle. Genetic ablation of TLR4 led to significantly increased muscle force generation in dystrophic diaphragm muscle, which was associated with improved histopathology including decreased fibrosis, as well as reduced pro-inflammatory gene expression and macrophage infiltration. TLR4 ablation in mdx mice also altered the phenotype of muscle macrophages by inducing a shift toward a more anti-inflammatory (iNOS(neg) CD206(pos)) profile. In vitro experiments confirmed that lack of TLR4 is sufficient to influence macrophage activation status in response to classical polarizing stimuli such as IFN-gamma and IL-4. Finally, treatment of dystrophic mice with glycyrrhizin, an inhibitor of the endogenous TLR4 ligand, high mobility group box (HMGB1), also pointed to involvement of the HMGB1-TLR4 axis in promoting dystrophic diaphragm pathology. Taken together, our findings reveal TLR4 and the innate immune system as important players in the pathophysiology of DMD. Accordingly, targeting either TLR4 or its endogenous ligands may provide a new therapeutic strategy to slow disease progression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
277258
2-Methylbutane, anhydrous, ≥99%
C5H12
270342
2-Methylbutane, for HPLC, ≥99.5%
C5H12
59060
2-Methylbutane, analytical standard
C5H12
59070
2-Methylbutane, puriss. p.a., ≥99.5% (GC)
C5H12
15-2230
2-Methylbutane, SAJ first grade, ≥99.0%
C5H12
M32631
2-Methylbutane, ReagentPlus®, ≥99%
C5H12
320404
2-Methylbutane, ReagentPlus®, ≥99%
C5H12
Y0000433
Ammonium glycyrrhizate, European Pharmacopoeia (EP) Reference Standard
C42H62O16 · NH3
SAB2700788
Anti-EMR1 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1405756
Anti-EMR1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA052809 Anti-EMR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500764
Anti-EMR1, C-Terminal antibody produced in rabbit, affinity isolated antibody
AV38110
Anti-HMGB1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2701685
Anti-hmgb1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701809
Anti-hmgb1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2101049
Anti-HMGB1 antibody produced in rabbit, affinity isolated antibody
SAB4501401
Anti-HMGB1 antibody produced in rabbit, affinity isolated antibody
89458
Aphidicolin, analytical standard
C20H34O4
G0460000
Glycyrrhizate monoammonium, European Pharmacopoeia (EP) Reference Standard
C42H62O16 · NH3
91443
Glycyrrhizic acid ammonium salt, analytical standard, for HPLC
C42H62O16 · NH3
01250570
Glycyrrhizic acid ammonium salt, primary pharmaceutical reference standard
C42H62O16 · NH3
G2137
Glycyrrhizic acid ammonium salt from glycyrrhiza root (licorice), ≥70% (HPLC)
C42H65NO16
50531
Glycyrrhizic acid ammonium salt from glycyrrhiza root (licorice), ≥95.0% (NT)
C42H65NO16
SAB4700268 Monoclonal Anti-CD16 antibody produced in mouse, clone MEM-168, purified immunoglobulin, buffered aqueous solution
SAB4700269 Monoclonal Anti-CD16 antibody produced in mouse, clone LNK16, purified immunoglobulin, buffered aqueous solution
SAB4700274 Monoclonal Anti-CD16 antibody produced in mouse, clone 3G8, purified immunoglobulin, buffered aqueous solution
SAB4700276 Monoclonal Anti-CD16 antibody produced in mouse, clone MEM-154, purified immunoglobulin, buffered aqueous solution
H9537
Monoclonal Anti-HMG-1 antibody produced in mouse, clone HAP46.5, purified from hybridoma cell culture
WH0003146M8
Monoclonal Anti-HMGB1 antibody produced in mouse, clone 2F6, purified immunoglobulin, buffered aqueous solution
SAB1403925
Monoclonal Anti-HMGB1 antibody produced in mouse, clone 1D5, purified immunoglobulin, buffered aqueous solution