EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Mixed alkyl aryl phosphonate esters as quenched fluorescent activity-based probes for serine proteases.
PMID 25553959
Abstract
Activity-based probes (ABPs) are powerful tools for the analysis of active enzyme species in whole proteomes, cells or animals. Quenched fluorescent ABPs (qABPs) can be applied for real time imaging, allowing the visualization of dynamic enzyme activation by fluorescent microscopy. Unfortunately, qABPs are only available for a few enzymes. We here describe the design and synthesis of qABPs for serine proteases based on a phosphonate ester scaffold.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4034
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
C6H7NaO6
11140
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
C6H7NaO6
05097
(+)-Sodium L-ascorbate, analytical reference material
C6H7NaO6
00990
Acetyl chloride, puriss. p.a., ≥99.0% (T)
C2H3ClO
B18200
Benzyl carbamate, 99%
C8H9NO2
254185
Copper(I) bromide, 99.999% trace metals basis
BrCu
212865
Copper(I) bromide, 98%
BrCu
61163
Copper(I) bromide, purum p.a., ≥98.0% (RT)
BrCu
735906
Copper(I) bromide, beads, -10 mesh, 99.99% trace metals basis
BrCu
361941
Di-tert-butyl dicarbonate, ReagentPlus®, ≥99%
C10H18O5
205249
Di-tert-butyl dicarbonate, ReagentPlus®, 99%
C10H18O5
34660
Di-tert-butyl dicarbonate, ≥98.0% (GC)
C10H18O5
436267
Di-tert-butyl dicarbonate solution, 1.0 M in THF
C10H18O5
703753
Di-tert-butyl dicarbonate solution, 2 M in THF
C10H18O5
703737
Di-tert-butyl dicarbonate solution, 2 M in methylene chloride
C10H18O5
703745
Di-tert-butyl dicarbonate solution, 2 M in ethyl acetate
C10H18O5
445460
HATU, 97%
C10H15F6N6OP
746479
Lithium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
BrLi
429465
Lithium bromide, anhydrous, beads, −10 mesh, 99.999% trace metals basis
BrLi
449873
Lithium bromide, anhydrous, beads, −10 mesh, ≥99.9% trace metals basis
BrLi
229733
Lithium bromide, powder and chunks, ≥99.995% trace metals basis
BrLi
213225
Lithium bromide, ReagentPlus®, ≥99%
BrLi
62464
Lithium bromide, puriss. p.a., ≥99.0% (dried material, AT)
BrLi
62463
Lithium bromide, purum p.a., ≥98.0% (dried material, AT), powder, crystals or chunks
BrLi
411515
Lithium bromide solution, 54 wt. % in H2O
BrLi
678139
Lithium bromide solution, 1.5 M in THF
BrLi
PHR1279
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
C6H7NaO6
Y0000039
Sodium ascorbate, European Pharmacopoeia (EP) Reference Standard
C6H7NaO6
71360
Sodium carbonate decahydrate, puriss. p.a., ≥99.0% (T)
CNa2O3 · 10H2O
13414
Sodium carbonate decahydrate, puriss., meets analytical specification of Ph. Eur., BP, FCC, E500, 99.5-100.5% (ex dried subst.)
CNa2O3 · 10H2O