EMAIL THIS PAGE TO A FRIEND
FEBS letters
Divergent signalling pathways regulate lipopolysaccharide-induced eRNA expression in human monocytic THP1 cells.
PMID 25554418
Abstract
Recent studies have indicated that non-coding RNAs transcribed from enhancer regions are important regulators of enhancer function and gene expression. In this report, we have characterised the expression of six enhancer RNAs (eRNAs) induced in human monocytic THP1 cells following activation of the innate immune response by lipopolysaccharide (LPS). Specifically, we have demonstrated that LPS-induced expression of individual eRNAs is mediated through divergent intracellular signalling pathways that includes NF-κB and the mitogen activated protein kinases, extracellular regulated kinase-1/2 and p38.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T1452
TPCA-1, ≥95% (HPLC)
C12H10FN3O2S