EMAIL THIS PAGE TO A FRIEND
Journal of nuclear medicine : official publication, Society of Nuclear Medicine
ACR white paper targets thyroid incidentaloma reporting.
PMID 25556226
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5006
2-Fluoro-2-deoxy-D-glucose, glycosylation inhibitor, glucose analog
C6H11FO5