EMAIL THIS PAGE TO A FRIEND
International neurourology journal
Influence of Panax ginseng on Alpha-Adrenergic Receptor of Benign Prostatic Hyperplasia.
PMID 25558416
Abstract
Benign prostatic hyperplasia (BPH) is the most common prostate problem in older men. The present study aimed to investigate the inhibitory effect of Panax ginseng C.A. Meyer (P. ginseng) on a rat model of testosterone-induced BPH. The rats were divided into 3 groups (each group, n=10): control, testosterone-induced BPH (20 mg/kg, subcutaneous injection), and P. ginseng (200 mg/kg, orally) groups. After 4 weeks, all animals were sacrificed to examine the blood biochemical profiles, prostate volume, weight, histopathological changes, alpha-1D adrenergic receptor (Adra1d) mRNA expression, and epidermal growth factor receptor (EGFR) and B-cell CLL/lymphoma 2 (BCL2) protein expression. The group treated with P. ginseng showed significantly lesser prostate size and weight than the testosterone-induced BPH group. In addition, P. ginseng decreased the mRNA expression of Adra1d as well as the expression of EGFR and BCL2 in prostate tissue. These results suggest that P. ginseng may inhibit the alpha-1-adrenergic receptor to suppress the development of BPH.