EMAIL THIS PAGE TO A FRIEND
Journal of intellectual disability research : JIDR
Cognitive shifting and externalising problem behaviour in intellectual disability and autism spectrum disorder.
PMID 25559338
Abstract
Behavioural problems are frequently reported in residential care for people with an intellectual disability (ID) in particular when they are additionally diagnosed with autism spectrum disorder (ASD). There are indications that impairment in cognitive shifting may be associated with problem behaviour. The objectives of this study were (1) to examine the relationship of cognitive shifting and severity of ASD symptoms with externalising problem behaviour in individuals with ID, with and without ASD, and (2) to examine whether a diagnosis based on shifting impairment is more predictive of externalising problem behaviour than an ASD diagnosis. Participants consisted of adolescents and young adults with mild ID, with and without ASD (n = 41). Pearson intercorrelations were computed to explore the relationship between shifting impairment and severity of ASD symptoms on the one hand and ratings of externalising problem behaviour on the other hand. t-Tests were performed to analyse differences in externalising problem behaviour. Unlike ASD symptom severity, shifting scores were found to be associated with externalising problem behaviour, but only if shifting was measured using rating scales and not when using neuropsychological tasks. Externalising problem behaviour scores significantly differed when groups were classified according to shifting impairment (impaired vs. non-impaired) but not when they were classified according to ID and ASD diagnoses. It is proposed to use a cognition-based approach when analysing problem behaviour, thus concentrating not so much on ID and ASD diagnosis and their corresponding symptoms, but rather placing the focus on cognitive symptoms.