EMAIL THIS PAGE TO A FRIEND
Critical care medicine
Prehospital aspirin use is associated with reduced risk of acute respiratory distress syndrome in critically ill patients: a propensity-adjusted analysis.
PMID 25559436
Abstract
Platelet activation plays an active role in the pathogenesis of acute respiratory distress syndrome. In our prior study of 575 patients at high risk for acute respiratory distress syndrome, concurrent statin and aspirin use was associated with reduced acute respiratory distress syndrome. However, the largest study (n = 3,855) to date found no significant benefit of prehospital aspirin in a lower-risk population when adjusted for the propensity for aspirin use. We aimed to determine whether prehospital aspirin use is associated with decreased acute respiratory distress syndrome in patients at high risk for acute respiratory distress syndrome after adjusting for the propensity to receive aspirin. Secondary analysis of patients enrolled prospectively in the Validating Acute Lung Injury Markers for Diagnosis study. A total of 1,149 critically ill patients (≥40 years old) admitted to the medical or surgical ICUs of an academic tertiary care hospital including 575 previously reported patients as well as additional patients who were enrolled after completion of the prior statin and aspirin study. None. Of 1,149 patients, 368 (32%) developed acute respiratory distress syndrome during the first 4 ICU days and 287 (25%) patients had prehospital aspirin use. Patients with prehospital aspirin had significantly lower prevalence of acute respiratory distress syndrome (27% vs 34%; p=0.034). In a multivariable, propensity-adjusted analysis including age, gender, race, sepsis, and Acute Physiology and Chronic Health Evaluation score II, prehospital aspirin use was associated with a decreased risk of acute respiratory distress syndrome (odds ratio, 0.66; 95% CI, 0.46-0.94) in the entire cohort and in a subgroup of 725 patients with sepsis (odds ratio, 0.60; 95% CI, 0.41-0.90). In this selected cohort of critically ill patients, prehospital aspirin use was independently associated with a decreased risk of acute respiratory distress syndrome even after adjusting for the propensity of prehospital aspirin use. These findings support the need for prospective clinical trials to determine whether aspirin may be beneficial for the prevention of clinical acute respiratory distress syndrome.