EMAIL THIS PAGE TO A FRIEND
Scientific reports
Orientation in multi-layer chitosan hydrogel: morphology, mechanism, and design principle.
PMID 25559867
Abstract
Hydrogels with organized structure have attracted remarkable attentions for bio-related applications. Among the preparation of hierarchical hydrogel materials, fabrication of hydrogel with multi-layers is an important branch. Although the generation mechanism of layers had been fully discussed, sub-layer structure was not sufficiently studied. In this research, multi-layered chitosan hydrogel with oriented structure was constructed, and the formation mechanism of orientation was proposed, based on gelation behavior and entanglement of polymer chains in the hydrogel-solution system. Employing the layered-oriented characteristic, chitosan hydrogel materials with various shapes and structure can be designed and fabricated.