EMAIL THIS PAGE TO A FRIEND
Journal of cellular biochemistry
Cinnamaldehyde and nitric oxide attenuate advanced glycation end products-induced the Jak/STAT signaling in human renal tubular cells.
PMID 25561392
Abstract
Cinnamaldehyde is a major and a bioactive compound isolated from the leaves of Cinnamomum osmophloeum kaneh. It possesses anti-diabetic properties in vitro and in vivo and has anti-inflammatory and anti-cancer effects. To explore whether cinnamaldehyde was linked to altered advanced glycation end products (AGE)-mediated diabetic nephropathy, the molecular mechanisms of cinnamaldehyde responsible for inhibition of AGE-reduced nitric oxide (NO) bioactivity in human renal proximal tubular cells were examined. We found that raising the ambient AGE concentration causes a dose-dependent decrease in NO generation. Cinnamaldehyde significantly reverses AGE-inhibited NO generation and induces high levels of cGMP synthesis and PKG activation. Treatments with cinnamaldehyde, the NO donor S-nitroso-N-acetylpenicillamine, and the JAK2 inhibitor AG490 markedly attenuated AGE-inhibited NOS protein levels and NO generation. Moreover, AGE-induced the JAK2-STAT1/STAT3 activation, RAGE/p27(Kip1) /collagen IV protein levels, and cellular hypertrophy were reversed by cinnamaldehyde. The ability of cinnamaldehyde to suppress STAT activation was also verified by the observation that it significantly increased SCOS-3 protein level. These findings indicate for the first time that in the presence of cinnamaldehyde, the suppression of AGE-induced biological responses is probably mediated by inactivating the JAK2-STAT1/STAT3 cascade or activating the NO pathway.