EMAIL THIS PAGE TO A FRIEND
Nature communications
A highly efficient polysulfide mediator for lithium-sulfur batteries.
PMID 25562485
Abstract
The lithium-sulfur battery is receiving intense interest because its theoretical energy density exceeds that of lithium-ion batteries at much lower cost, but practical applications are still hindered by capacity decay caused by the polysulfide shuttle. Here we report a strategy to entrap polysulfides in the cathode that relies on a chemical process, whereby a host--manganese dioxide nanosheets serve as the prototype--reacts with initially formed lithium polysulfides to form surface-bound intermediates. These function as a redox shuttle to catenate and bind 'higher' polysulfides, and convert them on reduction to insoluble lithium sulfide via disproportionation. The sulfur/manganese dioxide nanosheet composite with 75 wt% sulfur exhibits a reversible capacity of 1,300 mA h g(-1) at moderate rates and a fade rate over 2,000 cycles of 0.036%/cycle, among the best reported to date. We furthermore show that this mechanism extends to graphene oxide and suggest it can be employed more widely.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
03378
Chloride atomic spectroscopy standard concentrate 10.00 g Cl-, 10.00 g/L, for 1 l standard solution, analytical standard
HCl
529664
Manganese(IV) oxide, ≥99.99% trace metals basis
MnO2
203750
Manganese(IV) oxide, 99.99% trace metals basis
MnO2
243442
Manganese(IV) oxide, ReagentPlus®, ≥99%
MnO2
310700
Manganese(IV) oxide, 10 μm, reagent grade, ≥90%
MnO2
217646
Manganese(IV) oxide, activated, ~85%, <10 μm
MnO2
775711
Manganese(IV) oxide, nanorod, diam. × L 5-30 nm × 80-100 nm, 99% trace metals basis (excludes potassium content)
MnO2
P2097
Potassium permanganate, meets USP testing specifications
KMnO4
399124
Potassium permanganate, ACS reagent, ≥99.0%, low in mercury
KMnO4
238511
Potassium permanganate, ≤150 μm particle size, 97%
KMnO4
207985
Potassium permanganate, 97%
KMnO4
60459
Potassium permanganate, BioUltra, ≥99.0% (RT)
KMnO4
60458
Potassium permanganate, puriss. p.a., ACS reagent, Hg ≤0.000005%, ≥99.0% (RT)
KMnO4
31404
Potassium permanganate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
KMnO4
13206
Potassium permanganate, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%
KMnO4
60467
Potassium permanganate, tested according to Ph.Eur.
KMnO4
HT1022
Potassium permanganate solution
KMnO4
S7026
Sodium thiosulfate
Na2O3S2
563188
Sodium thiosulfate, ≥99.99% trace metals basis
Na2O3S2
HT1005
Sodium thiosulfate solution, 2 g/dL in deionized water
Na2O3S2
36576
Sulfur, PESTANAL®, analytical standard
S
213292
Sulfur, 99.998% trace metals basis
S
344621
Sulfur, flakes, ≥99.99% trace metals basis
S
414980
Sulfur, powder, 99.98% trace metals basis
S
HT-S200
Sulfur, prilled, ≥99.99 trace metals basis
S
179728
Super-Hydride® solution, 1.0 M lithium triethylborohydride in THF
C6H16BLi
401757
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
C4H8O
186562
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
C4H8O
439215
Tetrahydrofuran, for HPLC, ≥99.9%, inhibitor-free
C4H8O
437638
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
C4H8O