EMAIL THIS PAGE TO A FRIEND
Nature communications
A lysine-rich motif in the phosphatidylserine receptor PSR-1 mediates recognition and removal of apoptotic cells.
PMID 25564762
Abstract
The conserved phosphatidylserine receptor (PSR) was first identified as a receptor for phosphatidylserine, an 'eat-me' signal exposed by apoptotic cells. However, several studies suggest that PSR may also act as an arginine demethylase, a lysyl hydroxylase, or an RNA-binding protein through its N-terminal JmjC domain. How PSR might execute drastically different biochemical activities, and whether they are physiologically significant, remain unclear. Here we report that a lysine-rich motif in the extracellular domain of PSR-1, the Caenorhabditis elegans PSR, mediates specific phosphatidylserine binding in vitro and clearance of apoptotic cells in vivo. This motif also mediates phosphatidylserine-induced oligomerization of PSR-1, suggesting a mechanism by which PSR-1 activates phagocytosis. Mutations in the phosphatidylserine-binding motif, but not in its Fe(II) binding site critical for the JmjC activity, abolish PSR-1 phagocytic function. Moreover, PSR-1 enriches and clusters around apoptotic cells during apoptosis. These results establish that PSR-1 is a conserved, phosphatidylserine-recognizing phagocyte receptor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RES1427A-A7
Ammonium sulfate, PharmaGrade, NF, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
H8N2O4S
A4418
Ammonium sulfate, for molecular biology, ≥99.0%
H8N2O4S
A6387
Ammonium sulfate, ReagentPlus®, ≥99.0%
H8N2O4S
A3920
Ammonium sulfate, plant cell culture tested, ≥99.0%
H8N2O4S
09978
Ammonium sulfate, BioUltra, ≥99.5% (T)
H8N2O4S
A2939
Ammonium sulfate, BioXtra, ≥99.0%
H8N2O4S
204501
Ammonium sulfate, 99.999% trace metals basis
H8N2O4S
59845
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
H8N2O4S
451975
Ammonium sulfate-14N2 solution, 99.99 atom % 14N, 40 wt. % in H2O
H814N2O4S
485411
Ammonium-14N2 sulfate, 99.99 atom % 14N
H814N2O4S
485306
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
H814N2O4S
608602
Ammonium-14N2,sulfate-16O4, 99.99 atom % 14N, 99.99 atom % 16O
H814N216O4S
02672
Calcium atomic spectroscopy standard concentrate 1.00 g Ca, 1.00 g/L, for 1L standard solution, analytical standard
CaCl2
21059
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
CaCl2
21049
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
CaCl2
F7250
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
C21H11NO5S
PHR1359
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
C10H17N3O6S
Y0000517
Glutathione, European Pharmacopoeia (EP) Reference Standard
C10H17N3O6S
76524
Glycine, certified reference material, TraceCERT®
C2H5NO2
50049
Glycine, BioUltra, ≥99.0% (NT)
C2H5NO2
50050
Glycine, ≥99.0% (NT)
C2H5NO2
50058
Glycine, tested according to Ph.Eur.
C2H5NO2
PHR1111
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
C2H5NO2
1295800
Glycine, United States Pharmacopeia (USP) Reference Standard
C2H5NO2
94119
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
C2H5NO2
G0450000
Glycine, European Pharmacopoeia (EP) Reference Standard
C2H5NO2
54461
HEPES, ≥99.0% (T)
C8H18N2O4S
PHR1428
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
C8H18N2O4S
82762
Methanol, analytical standard
CH4O
PHR1372
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
CH4O