EMAIL THIS PAGE TO A FRIEND
The Journal of general virology
Characterization of mAbs to chicken anemia virus and epitope mapping on its viral protein, VP1.
PMID 25568186
Abstract
Three (MoCAV/F2, MoCAV/F8 and MoCAV/F11) of four mouse mAbs established against the A2/76 strain of chicken anemia virus (CAV) showed neutralization activity. Immunoprecipitation showed a band at ~50 kDa in A2/76-infected cell lysates by neutralizing mAbs, corresponding to the 50 kDa capsid protein (VP1) of CAV, and the mAbs reacted with recombinant VP1 proteins expressed in Cos7 cells. MoCAV/F2 and MoCAV/F8 neutralized the 14 CAV strains tested, whereas MoCAV/F11 did not neutralize five of the strains, indicating distinct antigenic variation amongst the strains. In blocking immunofluorescence tests with the A2/76-infected cells, binding of MoCAV/F11 was not inhibited by the other mAbs. MoCAV/F2 inhibited the binding of MoCAV/F8 to the antigens and vice versa, suggesting that the two mAbs recognized the same epitope. However, mutations were found in different parts of VP1 of the escape mutants of each mAb: EsCAV/F2 (deletion of T89+A90), EsCAV/F8 (I261T) and EsCAV/F11 (E144G). Thus, the epitopes recognized by MoCAV/F2 and MoCAV/F8 seemed to be topographically close in the VP1 structure, suggesting that VP1 has at least two different neutralizing epitopes. However, MoCAV/F8 did not react with EsCAV/F2 or EsCAV/F8, suggesting that binding of MoCAV/F8 to the epitope requires coexistence of the epitope recognized by MoCAV/F2. In addition, MoCAV/F2, with a titre of 1 : 12 800 to the parent strain, neutralized EsCAV/F2 and EsCAV/F8 with low titres of 32 and 152, respectively. The similarity of the reactivity of MoCAV/F2 and MoCAV/F8 to VP1 may also suggest the existence of a single epitope recognized by these mAbs.