EMAIL THIS PAGE TO A FRIEND
Journal of drug targeting
Biodistribution and anti-tumor efficacy of intratumorally injected necrosis-avid theranostic agent radioiodinated hypericin in rodent tumor models.
PMID 25572455
Abstract
Hypericin has an excellent necrosis-specific targeting capacity; thus, we explored small-molecular tumor necrosis therapy (SMTNT) for inhibiting tumor growth in rodent tumor models. H22 and S180 tumor-bearing Kunming (KM) mice were intratumorally injected with (131)I-monoiodohypericin ((131)I-MIH) to investigate the biodistribution of (131)I-MIH as a function of time. Single-photon emission computed tomography (SPECT), autoradiography, fluorescence microscopy and hematoxylin and eosin (H&E) staining were performed to determine the intra-tumoral distribution of (131)I-MIH. A therapeutic evaluation study was also performed in the tumor-bearing KM mice using saline and a positive drug as controls. Gamma counting, SPECT images, autoradiography and fluorescence microscopy and H&E staining results revealed intense retention of (131)I-MIH in the necrotic tumor over 168 h and good in vivo stability of the agent. Therapy with a single dose of intra-tumoral administration of (131)I-MIH caused significant tumor growth delay. A histopathological analysis of the tumors and normal organs further validated the therapeutic efficacy and limited systemic toxicity of (131)I-MIH. The prolonged tumor retention and effective therapy indicated that (131)I-MIH may be a promising intratumorally injected SMTNT agent.