EMAIL THIS PAGE TO A FRIEND
Plant physiology
HIGHLY METHYL ESTERIFIED SEEDS is a pectin methyl esterase involved in embryo development.
PMID 25572606
Abstract
Homogalacturonan pectin domains are synthesized in a highly methyl-esterified form that later can be differentially demethyl esterified by pectin methyl esterase (PME) to strengthen or loosen plant cell walls that contain pectin, including seed coat mucilage, a specialized secondary cell wall of seed coat epidermal cells. As a means to identify the active PMEs in seed coat mucilage, we identified seven PMEs expressed during seed coat development. One of these, HIGHLY METHYL ESTERIFIED SEEDS (HMS), is abundant during mucilage secretion, peaking at 7 d postanthesis in both the seed coat and the embryo. We have determined that this gene is required for normal levels of PME activity and homogalacturonan methyl esterification in the seed. The hms-1 mutant displays altered embryo morphology and mucilage extrusion, both of which are a consequence of defects in embryo development. A significant decrease in the size of cells in the embryo suggests that the changes in embryo morphology are a consequence of lack of cell expansion. Progeny from a cross between hms-1 and the previously characterized PME inhibitor5 overexpression line suggest that HMS acts independently from other cell wall-modifying enzymes in the embryo. We propose that HMS is required for cell wall loosening in the embryo to facilitate cell expansion during the accumulation of storage reserves and that its role in the seed coat is masked by redundancy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H2131
1-Hexadecene, ≥98.5%
C16H32
N8285
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
C21H27N7O14P2
N8410
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
C21H27N7O14P2
N8535
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
C21H27N7O14P2
N7004
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
C21H27N7O14P2 · xH2O
N6522
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
C21H27N7O14P2 · xH2O
N1636
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
C21H27N7O14P2 · xH2O
N3014
β-Nicotinamide adenine dinucleotide hydrate, cell culture tested, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
C21H27N7O14P2 · xH2O
N7381
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1, ≥95% (HPLC)
C21H27N7O14P2 · xH2O
N1511
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
C21H27N7O14P2 · xH2O
43410
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
C21H27N7O14P2 · xH2O
N7132
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
C21H26LiN7O14P2
P9135
Pectin from citrus peel, Galacturonic acid ≥74.0 % (dried basis)
R2751
Ruthenium Red, Technical grade
H42Cl6N14O2Ru3
84071
Ruthenium Red, for microscopy, ≥85% (calc. on dry substance, AT)
H42Cl6N14O2Ru3