EMAIL THIS PAGE TO A FRIEND
International journal of immunopathology and pharmacology
In vitro action of sho-seiryu-to on allergen-exposed mononuclear cells.
PMID 25572740
Abstract
Although Sho-seiryu-to (SST), used as a traditional herbal (Kampo) medicine mainly in China and Korea, is shown to have immunomodulating potential, such as an anti-allergic one, its underlying mechanism has not been completely clarified. To partially address the issue, we explored its effects on allergen-exposed mononuclear cells. Male balb/c mice were intraperitoneally administered ovalbumin (OVA: 20 μg) plus alum or vehicle twice (Day 0 and Day 14). At Day 21, mice were sacrificed and splenocytes (mononuclear cells) were isolated and cultured in the presence or absence of OVA with or without SST. Thereafter, helper T-related cytokines in the culture supernatants were evaluated by means of ELISA. Protein level of interferon-γ was lower than 5.0 pg/mL in the supernatants from OVAx96 non-exposed or -exposed mononuclear cells in the presence or absence of OVA stimulation. On the other hand, SST induced the cytokine from both types of mononuclear cells in the presence (P < 0.05) or absence of OVA stimulation as compared to corresponding control. By contrast, interleukin (IL)-4 level tended to be decreased by SST in OVA-non-exposed mononuclear cells as did IL-13 in both non-exposed and exposed mononuclear cells as compared to vehicle. In conclusion, immunoregulating efficacy by SST on allergy-prone subjects may include, at least in part, restoring helper T balance mainly through hyperproduction of IFN-γ against mononuclear cells such as lymphocytes.