EMAIL THIS PAGE TO A FRIEND
Acta neurobiologiae experimentalis
The role of the GABA-A receptor of the adjacent intact dorsal root ganglion neurons in rats with neuropathic pain.
PMID 25576971
Abstract
We aim to investigate the changes of The γ-aminobutyric acid (GABA) signals in the adjacent intact dorsal root ganglion (DRG) and the contribution of these changes to the development and maintenance of neuropathic pain (NPP). After establishment of neuropathic pain model with the lumbar 5 spinal nerve ligation (L5 SNL), the GABA-evoked currents were recorded in the acutely dissociated L4 DRG neurons using whole-cell patch clamp. Moreover, Muscimol or Bicuculline were respectively topically injected to the L4 DRG at the time of nerve injury and post-operative 5 days (POD5). The pain thresholds of hind paw were recorded. We found that the incidence and amplitude of GABA currents significantly decreased in the large and medium L4 DRG neurons after SNL (P<0.05). Furthermore, in the L5 SNL rats, 20 μg Muscimol injected immediately after injury caused a long-lasting attenuation of mechanical allodynia (P<0.05), whereas the thermal hyperalgesia was not alleviated (P>0.05). 0.15 μg Bicuculline further decreased the pain thresholds (P<0.05) and reversed the pain-alleviating effects of Muscimol. However, at POD5, 20 μg Muscimol only exhibited transient alleviating effects (P<0.05). In conclusion, the decrease of inhibition signal mediated by GABA-A receptor in the adjacent intact DRG neurons may be crucial in the development of hyperalgesia in NPP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30181
Fluorescein sodium salt, analytical standard
C20H10Na2O5