EMAIL THIS PAGE TO A FRIEND
Food chemistry
A validated HPLC method for the analysis of herbal teas from three chemotypes of Brazilian Lippia alba.
PMID 25577093
Abstract
Infusions and decoctions of three chemotypes of Lippia alba (Mill.) N. E. Brown (Verbenaceae) were investigated for their quantitative profiles by HPLC-DAD-ESI-MS analyses. An RP-HPLC method was developed which permitted the quality control of the preparations. The correct choice of the column allowed the detailed characterization of the constituents in a total analysis time of 35 min. The HPLC method was accordingly validated for linearity range, LOD, LOQ, accuracy and precision. For the quantitative analysis the three major phytochemical groups were taken into consideration, namely iridoids, phenylpropanoids and flavonoids. Comparative quantitative analyses revealed significant differences among the chemotypes that should be taken into account in the uses of the herbal teas. The developed HPLC-UV assay proved to be an efficient and alternative method for the discrimination of the three chemotypes. This is the first report of detailed analysis of the chemical composition of the constituents of L. alba chemotypes' teas.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
281689
1,1-Diphenyl-2-picrylhydrazine, 97%
C18H13N5O6
D9132
2,2-Diphenyl-1-picrylhydrazyl
C18H12N5O6