EMAIL THIS PAGE TO A FRIEND
Molecular plant
Vanillin-bioconversion and bioengineering of the most popular plant flavor and its de novo biosynthesis in the vanilla orchid.
PMID 25578271
Abstract
In recent years, biotechnology-derived production of flavors and fragrances has expanded rapidly. The world's most popular flavor, vanillin, is no exception. This review outlines the current state of biotechnology-based vanillin synthesis with the use of ferulic acid, eugenol, and glucose as substrates and bacteria, fungi, and yeasts as microbial production hosts. The de novo biosynthetic pathway of vanillin in the vanilla orchid and the possible applied uses of this new knowledge in the biotechnology-derived and pod-based vanillin industries are also highlighted.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W310700
Vanillin, ≥97%, FCC, FG
C8H8O3