EMAIL THIS PAGE TO A FRIEND
Clinical & experimental metastasis
RASSF5A, a candidate tumor suppressor, is epigenetically inactivated in esophageal squamous cell carcinoma.
PMID 25579665
Abstract
As a result of alternative splicing and differential promoter usage, RASSF5 exists in at least three isoforms (RASSF5A-RASSF5C), which may play different roles in tumorigenesis. The present study was to detect the role of RASSF5A, B and C in esophageal squamous cell carcinoma (ESCC) and clarify the critical CpG sites of RASSF5A, in order to clarify more information on the role of RASSF5 with regard to the pathogenesis of ESCC. Frequent silencing of RASSF5A but not RASSF5B and RASSF5C were found in esophageal cancer cell lines and the silencing of RASSF5A may be reversed by 5-Aza-dC or TSA treatment. The aberrant CpG island 1 methylation of RASSF5A induces silencing of its expression in TE13 cell line. Decreased mRNA and protein expression of RASSF5A was observed in ESCC tumor tissues and was associated with RASSF5A CpG island 1 methylation status. Unlike RASSF5A, expression variation of RASSF5B and RASSF5C was not found in ESCC tissues. Aberrant promoter methylation of RASSF5C was also not found in ESCC. RASSF5A methylation and protein expression were independently associated with ESCC patients' survival. These data indicated that the inactivation of RASSF5A through CpG island 1 methylation may play an important role in ESCC carcinogenesis, RASSF5A may be a functional tumor suppressor and may serve as a prognostic biomarker for ESCC.