EMAIL THIS PAGE TO A FRIEND
Wound repair and regeneration : official publication of the Wound Healing Society [and] the European Tissue Repair Society
The role of SDF-1 in homing of human adipose-derived stem cells.
PMID 25581571
Abstract
One of the putative pathophysiological mechanisms of chronic wounds is a disturbed homing of stem cells. In this project, the stromal cell-derived factor 1 (SDF-1)/C-X-C chemokine receptor (CXCR) 4 and SDF-1/CXCR7 pathway were focused in human adipose-derived stem cells (ASCs). ASCs were incubated with acute (AWF) or chronic wound fluid (CWF) to analyze their effects by quantitative real-time polymerase chain reaction (SDF-1, CXCR4, CXCR7, TIMP3), enzyme-linked immunosorbent assay (SDF-1 in WFs and supernatant), and transwell migration assay with/without antagonization. Whereas SDF-1 amounted 73.5 pg/mL in AWF, it could not be detected in CWF. Incubation with AWF led to a significant enhancement (129.7 pg/mL vs. 95.5 pg/mL), whereas CWF resulted in a significant reduction (30 pg/mL vs. 95.5 pg/mL) of SDF-1 in ASC supernatant. The SDF-1 receptor CXCR7 was detected on ASCs. AWF but not CWF significantly induced ASC migration, which was inhibited by CXCR4 and CXCR7 antagonists. Expressions of SDF-1, CXCR4, and CXCR7 were significantly stimulated by AWF while TIMP3 expression was reduced. In conclusion, an uncontrolled inflammation in the chronic wound environment, indicated by a reduced SDF-1 expression, resulted in a decreased ASC migration. A disturbed SDF-1/CXCR4 as well as SDF-1/CXCR7 pathway seems to play an important role in the impaired healing of chronic wounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA049718 Anti-ACKR3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1303785
ANTI-CMKOR1 (C-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1303471
ANTI-CXCR7 (C-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2700377
Anti-CXCR7 (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700198
Anti-CXCR7 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4502446
Anti-CXCR7, C-Terminal antibody produced in rabbit, affinity isolated antibody
HPA017357
Anti-NT5E antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
450146
Calcium hydroxide, 99.995% trace metals basis
H2CaO2
G6639
Guanidine thiocyanate, ≥97% (titration)
CH5N3 · CHNS
50980
Guanidine thiocyanate, BioUltra, for molecular biology, ≥99.0% (AT)
CH5N3 · CHNS
50981
Guanidine thiocyanate, BioReagent, for molecular biology, ≥99%
CH5N3 · CHNS
G9277
Guanidine thiocyanate, for molecular biology, ≥99%
CH5N3 · CHNS
WH0004907M1
Monoclonal Anti-NT5E antibody produced in mouse, clone 4C4-2B5, purified immunoglobulin, buffered aqueous solution
SAB5300408 Monoclonal Anti-NT5E antibody produced in mouse, clone 1D7, ascites fluid
SAB1404149
Monoclonal Anti-NT5E, (C-terminal) antibody produced in mouse, clone 4D3-2C10, purified immunoglobulin, buffered aqueous solution
SAB1402533
Monoclonal Anti-NTE, (C-terminal) antibody produced in mouse, clone 3D10, purified immunoglobulin, buffered aqueous solution