EMAIL THIS PAGE TO A FRIEND
Alcoholism, clinical and experimental research
N-acetylcysteine inhibits the up-regulation of mitochondrial biogenesis genes in livers from rats fed ethanol chronically.
PMID 25581647
Abstract
Chronic ethanol (EtOH) administration to experimental animals induces hepatic oxidative stress and up-regulates mitochondrial biogenesis. The mechanisms by which chronic EtOH up-regulates mitochondrial biogenesis have not been fully explored. In this work, we hypothesized that oxidative stress is a factor that triggers mitochondrial biogenesis after chronic EtOH feeding. If our hypothesis is correct, co-administration of antioxidants should prevent up-regulation of mitochondrial biogenesis genes. Rats were fed an EtOH-containing diet intragastrically by total enteral nutrition for 150xa0days, in the absence or presence of the antioxidant N-acetylcysteine (NAC) at 1.7xa0g/kg/d; control rats were administered isocaloric diets where carbohydrates substituted for EtOH calories. EtOH administration significantly increased hepatic oxidative stress, evidenced as decreased liver total glutathione and reduced glutathione/glutathione disulfide ratio. These effects were inhibited by co-administration of EtOH and NAC. Chronic EtOH increased the expression of mitochondrial biogenesis genes including peroxisome proliferator-activated receptor gamma-coactivator-1 alpha and mitochondrial transcription factor A, and mitochondrial DNA; co-administration of EtOH and NAC prevented these effects. Chronic EtOH administration was associated with decreased mitochondrial mass, inactivation and depletion of mitochondrial complex I and complex IV, and increased hepatic mitochondrial oxidative damage, effects that were not prevented by NAC. These results suggest that oxidative stress caused by chronic EtOH triggered the up-regulation of mitochondrial biogenesis genes in rat liver, because an antioxidant such as NAC prevented both effects. Because NAC did not prevent liver mitochondrial oxidative damage, extra-mitochondrial effects of reactive oxygen species may regulate mitochondrial biogenesis. In spite of the induction of hepatic mitochondrial biogenesis genes by chronic EtOH, mitochondrial mass and function decreased probably in association with mitochondrial oxidative damage. These results also predict that the effectiveness of NAC as an antioxidant therapy for chronic alcoholism will be limited by its limited antioxidant effects in mitochondria, and its inhibitory effect on mitochondrial biogenesis.