EMAIL THIS PAGE TO A FRIEND
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie
Four-year to seven-year outcomes of advanced surface ablation with excimer laser for high myopia.
PMID 25582070
Abstract
We aimed to evaluate and compare outcomes after photorefractive keratectomy with cooling (cPRK) and laser-assisted subepithelial keratectomy (LASEK) for high myopia. This was a retrospective, single-masked follow-up study of patients treated for myopia between 2007 and 2009 with cPRK or LASEK, using a high-frequency flying-spot excimer laser with eye-tracker (MEL80; Carl Zeiss, Jena, Germany). One eye of each patient was randomly chosen for analysis. Re-treated eyes were excluded. Forty-six cPRK patients and 35 LASEK patients were included. Spherical equivalent averaged -7.69 ± 1.47 diopters (D) in cPRK eyes and -7.98 ± 2.06 D in LASEK eyes (p = 0.31) before surgery. The average follow-up time was 4.6 years in cPRK patients and 6.0 years in LASEK patients (p < 0.05). At final follow-up, no cPRK eyes and one LASEK eye (p = 0.46) had lost two lines of corrected distance visual acuity (CDVA). No eyes had significant haze at final follow-up, although trace haze was found in four cPRK eyes and six LASEK eyes (p = 0.44). However, at 6 weeks after surgery, zero cPRK eyes and nine LASEK eyes (p < 0.05) had significant haze. At final follow-up, 63 % of cPRK eyes and 35 % of LASEK eyes (p = 0.17) were within ±1.0 D of intended refraction. Finally, 100 % of cPRK patients and 92 % of LASEK patients (p = 0.87) were satisfied or very satisfied with the surgery at final follow-up. cPRK and LASEK seemed safe and with high patient satisfaction 4 to 7 years after surgery for high myopia. However, cPRK was more effective than LASEK in reducing initial significant corneal haze.
Related Materials