EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
On the use of aptamer microarrays as a platform for the exploration of human prothrombin/thrombin conversion.
PMID 25582304
Abstract
Microarrays are particular biosensors with multiple grafted probes that are generally used for parallel and simultaneous detection of various targets. In this study, we used microarrays with aptamer probes in order to follow up the different biomolecular interactions of a single enzyme, the thrombin protein, involved in the complex coagulation cascade. More precisely, thanks to label-free surface plasmon resonance imaging, we were able to monitor in real time an important step in the firing of the coagulation cascade in situ-the enzymatic transformation of prothrombin into thrombin, catalyzed by factor Xa. We were also able to appraise the influence of other biochemical factors and their corresponding inhibiting or enhancing behaviors on thrombin activation. Our study opens the door for the development of a complete microarray-based platform not only for the whole coagulation cascade analysis but also for novel drug screening assays in pharmacology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
674427
11-Mercaptoundecanoic acid, 98%
C11H22O2S
450561
11-Mercaptoundecanoic acid, 95%
C11H22O2S