EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Direct sample introduction gas chromatography and mass spectrometry for the determination of phthalate esters in cleaning products.
PMID 25582486
Abstract
A method using direct sample introduction (DSI) coupled to gas chromatography-mass spectrometry (GC-MS) is developed for the determination of six phthalate esters (dimethyl, diethyl, dibutyl, butylbenzyl, diethylhexyl and dioctyl phthalate) in cleaning products. The different variables involved in the DSI step, including venting time and temperature, vaporisation time and temperature, injector heating temperature and gas flow rate and pressure, were evaluated and optimised using Taguchi orthogonal arrays. The proposed method, using calibration against methanolic standards, showed good linearity in the 0.05-15 μg g(-1) range and good repeatability, with RSD values ranging from 3.5% to 5.7%. Quantification limits between 0.010 and 0.041 μg g(-1), depending on the compound, were attained, while recovery assays provided values from 83% to 115%. Twenty-seven cleaning products were analysed using the DSI-GC-MS method, being four phthalates (dimethyl, diethyl, dibutyl and diethylhexyl phthalate) found in fourteen of them at concentration levels in the 0.1-21 μg g(-1) range. Compared with the most common GC injection technique, which uses the split/splitless injector, the proposed DSI procedure provided larger peak areas and lower detection limits, as result of the greater injected volume and reduction in noise.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07671
Anthracene, certified reference material, TraceCERT®
C14H10
31581
Anthracene, analytical standard
C14H10
48567
Anthracene, analytical standard
C14H10
10580
Anthracene, suitable for scintillation, ≥99.0% (GC)
C14H10
331481
Anthracene, zone-refined, ≥99%
C14H10
141062
Anthracene, ReagentPlus®, 99%
C14H10
A89200
Anthracene, reagent grade, 97%
C14H10
694959
Anthracene, sublimed grade, ≥99%
C14H10
ERA-010
Anthracene, vial of 250 mg, analytical standard
C14H10
40076
Anthracene solution, certified reference material, 1000 μg/mL in acetone
C14H10
48647
Anthracene solution, certified reference material, 200 μg/mL in methanol
C14H10
67261
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®
C24H38O4
36735
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
C24H38O4
80030
Bis(2-ethylhexyl) phthalate, Selectophore
C24H38O4
89353
Density Standard 1191 kg/m3, H&D Fitzgerald Ltd. Quality
C10H10O4
88173
Di-n-octyl phthalate, certified reference material, TraceCERT®
C24H38O4
31301
Di-n-octyl phthalate, analytical standard
C24H38O4
80153
Di-n-octyl phthalate, ≥98.0% (GC)
C24H38O4
48560-U
Di-n-octyl phthalate, analytical standard
C24H38O4
18281
Dibutyl phthalate, certified reference material, TraceCERT®
C16H22O4
D1010000
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
C16H22O4
80100
Dibutyl phthalate, Selectophore
C16H22O4
36736
Dibutyl phthalate, PESTANAL®, analytical standard
C16H22O4
36737
Diethyl phthalate, PESTANAL®, analytical standard
C12H14O4
53008
Diethyl phthalate, PESTANAL®, analytical standard
C12H14O4
524972
Diethyl phthalate, 99.5%
C12H14O4
W512206
Diethyl phthalate, ≥99%
C12H14O4
D1410000
Diethyl phthalate, European Pharmacopoeia (EP) Reference Standard
C12H14O4
PHR1476
Diethyl Phthalate, Pharmaceutical Secondary Standard; Certified Reference Material
C12H14O4
32490
Diethyl pyrocarbonate, purum, ≥97.0% (NT)
C6H10O5