EMAIL THIS PAGE TO A FRIEND
Developmental cell
Regulating the timing of CENP-A nucleosome assembly by phosphorylation.
PMID 25584791
Abstract
In this issue of Developmental Cell, Yu et al. (2015) demonstrate that CENP-A phosphorylation by CDK1 inhibits its association with the chaperone protein HJURP and that the removal of this modification at mitotic exit is a key regulatory event that controls the timing of new CENP-A nucleosome formation at centromeres.