EMAIL THIS PAGE TO A FRIEND
Journal of molecular biology
Proteomic characterization of the nucleolar linker histone H1 interaction network.
PMID 25584861
Abstract
To investigate the relationship between linker histone H1 and protein-protein interactions in the nucleolus, we used biochemical and proteomics approaches to characterize nucleoli purified from cultured human and mouse cells. Mass spectrometry identified 175 proteins in human T cell nucleolar extracts that bound to Sepharose-immobilized H1 in vitro. Gene ontology analysis found significant enrichment for H1 binding proteins with functions related to nucleolar chromatin structure and RNA polymerase I transcription regulation, rRNA processing, and mRNA splicing. Consistent with the affinity binding results, H1 existed in large (400 to >650kDa) macromolecular complexes in human T cell nucleolar extracts. To complement the biochemical experiments, we investigated the effects of in vivo H1 depletion on protein content and structural integrity of the nucleolus using the H1 triple isoform knockout (H1ΔTKO) mouse embryonic stem cell (mESC) model system. Proteomic profiling of purified wild-type mESC nucleoli identified a total of 613 proteins, only ~60% of which were detected in the H1 mutant nucleoli. Within the affected group, spectral counting analysis quantitated 135 specific nucleolar proteins whose levels were significantly altered in H1ΔTKO mESC. Importantly, the functions of the affected proteins in mESC closely overlapped with those of the human T cell nucleolar H1 binding proteins. Immunofluorescence microscopy of intact H1ΔTKO mESC demonstrated both a loss of nucleolar RNA content and altered nucleolar morphology resulting from in vivo H1 depletion. We conclude that H1 organizes and maintains an extensive protein-protein interaction network in the nucleolus required for nucleolar structure and integrity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2500246
Anti-CLIC4 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2701750
Anti-clic4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA008019
Anti-CLIC4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV35222
Anti-CLIC4 antibody produced in rabbit, IgG fraction of antiserum
SAB1303876 ANTI-HIST1H1B (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
AV40583
Anti-NCL (AB1) antibody produced in rabbit, affinity isolated antibody
AV40584
Anti-NCL (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1409341
Anti-NCL antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA023981
Anti-NCL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410823
Anti-NCL antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
N2662
Anti-Nucleolin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1401336
Anti-RLN1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2106002
Anti-RLN1 antibody produced in rabbit, affinity isolated antibody
SAB1401521
Anti-TUBA1B antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1407132
Anti-VPS45 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA027425 Anti-VPS45 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA027441
Anti-VPS45 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
SAB2106973
Anti-VPS45 antibody produced in rabbit, affinity isolated antibody
SAB2501108
Anti-VPS45, C-Terminal antibody produced in goat, affinity isolated antibody, buffered aqueous solution
WH0010376M1
Monoclonal Anti-K-ALPHA-1 antibody produced in mouse, clone 4D1, purified immunoglobulin, buffered aqueous solution
WH0006013M1
Monoclonal Anti-RLN1 antibody produced in mouse, clone 1H6, purified immunoglobulin, buffered aqueous solution
SAB1404816
Monoclonal Anti-TUBA1B antibody produced in mouse, clone 2E8, purified immunoglobulin, buffered aqueous solution
P9172
Pyronin Y, for NA electrophoresis
C17H19ClN2O
83200
Pyronin Y, for microscopy (Bot., Fl., Hist.)
C17H19ClN2O
213519
Pyronin Y, certified by the Biological Stain Commission, Dye content 50 %
C17H19ClN2O