EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
New abietane-type diterpene glycosides from the roots of Tripterygium wilfordii.
PMID 25588600
Abstract
Two new abietane diterpene glycosides, wilfordosides A (1) and B (2), were isolated from the roots of Tripterygium wilfordii. The structures of compounds 1 and 2 were established using spectroscopic methods including extensive 1D and 2D NMR analysis, in combination with chemical reactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H1637
cis-4-Hydroxy-L-proline, collagen synthesis inhibitor
C5H9NO3