EMAIL THIS PAGE TO A FRIEND
Molecular cancer therapeutics
Mechanism of Oncogenic Signal Activation by the Novel Fusion Kinase FGFR3-BAIAP2L1.
PMID 25589496
Abstract
Recent cancer genome profiling studies have identified many novel genetic alterations, including rearrangements of genes encoding FGFR family members. However, most fusion genes are not functionally characterized, and their potentials in targeted therapy are unclear. We investigated a recently discovered gene fusion between FGFR3 and BAI1-associated protein 2-like 1 (BAIAP2L1). We identified 4 patients with bladder cancer and 2 patients with lung cancer harboring the FGFR3-BAIAP2L1 fusion through PCR and FISH assay screens. To investigate the oncogenic potential of the fusion gene, we established an FGFR3-BAIAP2L1 transfectant with Rat-2 fibroblast cells (Rat-2_F3-B). The FGFR3-BAIAP2L1 fusion had transforming activity in Rat2 cells, and Rat-2_F3-B cells were highly tumorigenic in mice. Rat-2_F3-B cells showed in vitro and in vivo sensitivity in the selective FGFR inhibitor CH5183284/Debio 1347, indicating that FGFR3 kinase activity is critical for tumorigenesis. Gene signature analysis revealed that FGFR3-BAIAP2L1 activates growth signals, such as the MAPK pathway, and inhibits tumor-suppressive signals, such as the p53, RB1, and CDKN2A pathways. We also established Rat-2_F3-B-ΔBAR cells expressing an FGFR3-BAIAP2L1 variant lacking the Bin-Amphiphysin-Rvs (BAR) dimerization domain of BAIAP2L1, which exhibited decreased tumorigenic activity, FGFR3 phosphorylation, and F3-B-ΔBAR dimerization, compared with Rat-2_F3-B cells. Collectively, these data suggest that constitutive dimerization through the BAR domain promotes constitutive FGFR3 kinase activation and is essential for its potent oncogenic activity.